Skip to Content
Merck
All Photos(1)

Key Documents

Q2125

Supelco

Quinaldine

analytical standard, ≥90% (GC)

Synonym(s):

2-Methylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
Beilstein:
110309
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.612 (lit.)

bp

105-107 °C/10 mmHg (lit.)
248 °C (lit.)

mp

−9-−3 °C (lit.)

density

1.058 g/mL at 25 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ccc2ccccc2n1

InChI

1S/C10H9N/c1-8-6-7-9-4-2-3-5-10(9)11-8/h2-7H,1H3

InChI key

SMUQFGGVLNAIOZ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Ferri et al.
Journal of the American Chemical Society, 123(48), 12074-12084 (2001-11-29)
An in situ attenuated total reflection study of the chiral solid-liquid interface created by cinchonidine adsorption on a Pt/Al(2)O(3) model catalyst is presented. Experiments were performed in the presence of dissolved hydrogen, that is under conditions used for the heterogeneous
Miguel Rubio et al.
Molecules (Basel, Switzerland), 15(11), 7732-7741 (2010-11-03)
The hydrogenation of 2-methylquinoline with Ir catalysts based on chiral phosphine-phosphites has been investigated. It has been observed that the reaction is very sensitive to the nature of the ligand. Optimization of the catalyst, allowed by the highly modular structure
Yongmei Li et al.
Journal of hazardous materials, 173(1-3), 151-158 (2009-09-08)
The anaerobic degradation of quinoline, isoquinoline and 2-methylquinoline was investigated under nitrate-reducing conditions with acclimated activated sludge. Quinoline was completely transformed during degradation with an optimum COD/NO(3)-N ratio of 7. Isoquinoline and 2-methylquinoline were also completely transformed; however, nitrate consumption
J Ortuño et al.
Fish & shellfish immunology, 12(1), 49-59 (2002-02-28)
Anaesthesia may depress the immune system in mammals, but there is no available information on this topic in fish. In the present work, four anaesthetics that are used in aquaculture, MS222 (0 19 mM), benzocaine (0.21 mM), 2-phenoxyethanol (16 mM)
Robert M Gengan et al.
Molecules (Basel, Switzerland), 15(5), 3171-3178 (2010-07-27)
An efficient synthesis of a methyl derivative of the indoloquinoline alkaloid cryptosanguinolentine based on microwave-assisted reactions is described. The microwave-assisted synthesis of an intermediate 4-hydroxy-2-methylquinoline yielded 86% of the desired product and other intermediates prepared yielded high % of products

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service