PHR3494
1,2-Dichloropropane
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1,2-Dichloropropane, Propylene dichloride
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
vapor density
3.89 (vs air)
vapor pressure
40 mmHg ( 19.4 °C)
form
liquid
autoignition temp.
1035 °F
expl. lim.
14.5 %
packaging
pkg of 1 mL
refractive index
n20/D 1.439 (lit.)
bp
95-96 °C (lit.)
mp
−100 °C (lit.)
density
1.156 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
2-30°C
SMILES string
CC(Cl)CCl
InChI
1S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3
InChI key
KNKRKFALVUDBJE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
59.0 °F
Flash Point(C)
15 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service