Skip to Content
Merck
All Photos(1)

Key Documents

PHR1874

Supelco

Atorvastatin Related Compound I

pharmaceutical secondary standard, certified reference material

Synonym(s):

tert-Butyl (4R,6R)-6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1-pyrrolyl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetate, tert-Butyl 2-((4R,6R)-6-{2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl}-2,2-dimethyl-1,3-dioxan-4-yl)acetate, (4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetic acid 1,1-dimethylethyl ester, tert-Butyl 2-[(4R,6R)-6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H47FN2O5
CAS Number:
Molecular Weight:
654.81
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1044593

API family

atorvastatin

CofA

current certificate can be downloaded

packaging

pkg of 30 mg

mp

144-148 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1c(C(=O)Nc2ccccc2)c(-c3ccccc3)c(-c4ccc(F)cc4)n1CC[C@@H]5C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O5

InChI

1S/C40H47FN2O5/c1-26(2)36-35(38(45)42-30-16-12-9-13-17-30)34(27-14-10-8-11-15-27)37(28-18-20-29(41)21-19-28)43(36)23-22-31-24-32(47-40(6,7)46-31)25-33(44)48-39(3,4)5/h8-21,26,31-32H,22-25H2,1-7H3,(H,42,45)/t31-,32-/m1/s1

InChI key

NPPZOMYSGNZDKY-ROJLCIKYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This material is an impurity of atorvastatin. Atorvastatin is a synthetic HMG-CoA reductase inhibitor which lowers plasma cholesterol levels by inhibiting endogenous cholesterol synthesis.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Atorvastatin may be used as a pharmaceutical reference standard for the determination of the analyte in drug dosage forms by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB1040 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An HPLC method for the determination of atorvastatin and its impurities in bulk drug and tablets
Erturk S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 33(5), 1017-1023 (2003)
HPLC methods for the determination of simvastatin and atorvastatin
Novakova L, et al.
TrAC, Trends in Analytical Chemistry, 27(4), 352-367 (2008)
Atorvastatin associated liver disease
Clarke, AT and Mills, PR
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver, 38(10), 772-777 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service