Skip to Content
Merck
All Photos(1)

Key Documents

PHR1682

Supelco

Baclofen

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(±)-Baclofen, (±)-β-(Aminomethyl)-4-chlorobenzenepropanoic acid, Lioresal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClNO2
CAS Number:
Molecular Weight:
213.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 28
traceable to Ph. Eur. B0200000
traceable to USP 1048200

API family

baclofen

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

ClC1=CC=C(C(CN)CC(O)=O)C=C1

InChI

1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)

InChI key

KPYSYYIEGFHWSV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Baclofen is a structural analog of γ-aminobutyric acid (GABA) and an antispastic agent commonly used for the treatment of spasticity as a result of spinal cord or brain injury.[1]

Application

Baclofen may be used as a pharmaceutical secondary reference standard for the determination of the analyte in plasma samples by liquid chromatography tandem mass spectrometry[1] and tablet formulations by UV spectroscopy,[2] respectively.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

GABAB receptor agonist; skeletal muscle relaxant; antispastic agent.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6298 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development and validation of baclofen mouth dissolving tablets by UV spectroscopy
Islam T, et al.
European Journal of Applied Science, 5(1), 7-11 (2013)
Liquid chromatography--tandem mass spectrometry determination of baclofen in various biological samples and application to a pharmacokinetic study
Kim TH, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 938, 43-50 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service