Skip to Content
Merck
All Photos(1)

Key Documents

PHR1388

Supelco

Adapalene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Adapalene, 6-(4-Methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic acid, 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid, CD-271, Differin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28O3
CAS Number:
Molecular Weight:
412.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001121
traceable to USP 1011709

API family

adapalene

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccc(cc1C23C[C@H]4C[C@H](C[C@H](C4)C2)C3)-c5ccc6cc(ccc6c5)C(O)=O

InChI

1S/C28H28O3/c1-31-26-7-6-23(21-2-3-22-12-24(27(29)30)5-4-20(22)11-21)13-25(26)28-14-17-8-18(15-28)10-19(9-17)16-28/h2-7,11-13,17-19H,8-10,14-16H2,1H3,(H,29,30)/t17-,18+,19-,28-

InChI key

LZCDAPDGXCYOEH-AADAIPAGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adapalene is a third-generation synthetic retinoid and a highly lipophilic compound derived from naphthoic acid. It is widely used in the treatment of acne.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Adapalene may be used as a pharmaceutical reference standard for the quantification of the analyte in topical gel formulations using high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Retinoic acid analogue that is a RARβ and RARγ agonist (AC50 values are 2.2, 9.3, 22 and > 1000 nM for RARβ, RARγ, RARα and RXRα receptors respectively). Inhibits proliferation and induces apoptosis in colorectal cancer cells in vitro. Displays comedolytic activity. Its unique pharmacological properties make it superior to other retinoids for the treatment of acne.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3530 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel stability-indicating RP-HPLC method for the simultaneous estimation of clindamycin phosphate and adapalene along with preservatives in topical gel formulations
Modi.BP and Shah.JN
Scientia Pharmaceutica, 82(4), 799-814 (2014)
A simple HPLC-DAD method for determination of adapalene in topical gel formulation
Martins AL, et al.
Journal of Chromatographic Science, 49(10), 796-800 (2011)
B Shroot et al.
Journal of the American Academy of Dermatology, 36(6 Pt 2), S96-103 (1997-06-01)
Retinoid research in the field of dermatology has been influenced by the clinical success of topical tretinoin and oral isotretinoin in the treatment of acne, and by the discovery of high-affinity binding proteins for retinoic acid mediating its action and
R N Brogden et al.
Drugs, 53(3), 511-519 (1997-03-01)
Adapalene, a naphthoic acid derivative with retinoid-like activity, is used for the topical treatment of mild to moderate acne vulgaris. It binds to retinoic acid receptors found predominantly in the terminal differentiation zone of epidermis and is more active than
J C English et al.
Cutis, 63(4), 227-230 (1999-05-06)
Darier's disease is an autosomal dominant disorder of keratinization that can produce unique clinical manifestations. Oral and topical retinoid products have been the mainstay therapy for this population, but local as well as systemic side effects often limit their effectiveness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service