Skip to Content
Merck
All Photos(1)

Key Documents

PHR1338

Supelco

Triclosan

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Triclosan, 5-Chloro-2-(2,4-dichlorophenoxy)phenol, Irgasan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3O2
CAS Number:
Molecular Weight:
289.54
Beilstein:
2057142
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1682206

API family

triclosan

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl

InChI

1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H

InChI key

XEFQLINVKFYRCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.
Triclosan is a non-ionic broad spectrum antibacterial and antifungal agent, used in personal care products such as antiseptic soaps, toothpastes, fabrics and plastics.

Application

A component of cefsulodin-irgasan-novobiocin selection medium for Yersinia, a human pathogen that may contaminate animal-source food products.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Triclosan may be used as a pharmaceutical reference standard for the determination of the analyte in waste water samples and food stuffs by various chromatography methods.

Biochem/physiol Actions

It is an inhibitor of the enoyl-ACP (acyl-carrier protein) reductase component of type II fatty acid synthase (FAS-II) in bacteria and Plasmodium. It also inhibits mammalian fatty acid synthase (FASN), and may have anticarcinogenic activity.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9502 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of triclosan in foodstuffs
Sanches-Silva A, et al.
Journal of Separation Science, 28(1), 65-72 (2005)
Triclosan: applications and safety
Bhargava HN and Leonard PA
American Journal of Infection Control, 24(3), 209-218 (1996)
Markus K Diener et al.
Lancet (London, England), 384(9938), 142-152 (2014-04-11)
Postoperative surgical site infections are one of the most frequent complications after open abdominal surgery, and triclosan-coated sutures were developed to reduce their occurrence. The aim of the PROUD trial was to obtain reliable data for the effectiveness of triclosan-coated
Andrea B Dann et al.
Journal of applied toxicology : JAT, 31(4), 285-311 (2011-04-05)
Triclosan [5-chloro-2-(2,4-dichlorophenoxy)phenol; TCS] is a broad spectrum antibacterial agent used in personal care, veterinary, industrial and household products. TCS is commonly detected in aquatic ecosystems, as it is only partially removed during the wastewater treatment process. Sorption, biodegradation and photolytic
Anthony Blinkhorn et al.
British dental journal, 207(3), 117-125 (2009-08-08)
Dental caries and periodontal disease are the most common oral conditions experienced by adults today. The treatment of these diseases by the dental team can only be performed when patients attend dental practices. There is recognition that the preventive measures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service