Skip to Content
Merck
All Photos(1)

Documents

PHR1207

Supelco

Methyl Ethyl Ketone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Butanone, Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1430101

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

960 °F

expl. lim.

10.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 2-Butanone (Methyl ethyl ketone, MEK), an aliphatic ketone, is considered as an aprotic and protophilic solvent. Its ability as an anticonvulsive agent in lithium-pilocarpine-induced status epilepticus in rats is reported.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0146 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Issues in Pharmacology, Pharmacy, Drug Research, and Drug Innovation: 2011 Edition (2012)
Yang Cao et al.
British journal of haematology, 168(5), 701-707 (2014-11-06)
CXCR4(WHIM) frameshift and nonsense mutations follow MYD88(L265P) as the most common somatic variants in Waldenström Macroglobulinaemia (WM), and impact clinical presentation and ibrutinib response. While the nonsense (CXCR4(S338X) ) mutation has been investigated, little is known about CXCR4 frameshift (CXCR4(FS)
Federico Mele et al.
Nature communications, 6, 6431-6431 (2015-03-17)
T helper (TH) cell polarization during priming is modulated by a number of signals, but whether polarization to a given phenotype also influences recall responses of memory TH cells is relatively unknown. Here we show that miR-181a is selectively induced
Brinda Alagesan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(2), 396-404 (2014-10-29)
Improved therapeutic approaches are needed for the treatment of pancreatic ductal adenocarcinoma (PDAC). As dual MEK and PI3K inhibition is presently being used in clinical trials for patients with PDAC, we sought to test the efficacy of combined targeting of
Inna V Fedorenko et al.
Proteomics, 15(2-3), 327-339 (2014-10-24)
Basal and kinase inhibitor driven adaptive signaling has been examined in a panel of melanoma cell lines using phosphoproteomics in conjunction with pathway analysis. A considerable divergence in the spectrum of tyrosine-phosphorylated peptides was noted at the cell line level.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service