Skip to Content
Merck
All Photos(1)

Key Documents

PHL82226

Cucurbitacin B

phyproof® Reference Substance

Synonym(s):

Amarine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H46O8
CAS Number:
Molecular Weight:
558.70
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1[C@@H]([C@@]2([C@]([C@H]3[C@]([C@@H]4C[C@@H](C(=O)C(C4=CC3)(C)C)O)(C(=O)C2)C)(C1)C)C)[C@](O)(C)C(=O)\C=C\C(OC(=O)C)(C)C

InChI

1S/C32H46O8/c1-17(33)40-27(2,3)13-12-23(36)32(9,39)25-21(35)15-29(6)22-11-10-18-19(14-20(34)26(38)28(18,4)5)31(22,8)24(37)16-30(25,29)7/h10,12-13,19-22,25,34-35,39H,11,14-16H2,1-9H3/b13-12+/t19-,20+,21-,22+,25+,29+,30-,31+,32+/m1/s1

InChI key

IXQKXEUSCPEQRD-DKRGWESNSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinia R Dwijayanti et al.
Phytotherapy research : PTR, 34(6), 1338-1346 (2019-12-18)
Bitter melon (Momordica charantia L.) is a vegetable and has been used as traditional medicine. Recently, we reported that bitter melon fruit extracts and its ethyl acetate (EtOAc)-soluble fraction markedly suppressed the expression of proinflammatory genes, including the inducible nitric
Sirinapha Klungsaeng et al.
The American journal of Chinese medicine, 48(6), 1475-1489 (2020-09-11)
Inadequate responses to traditional chemotherapeutic agents in cholangiocarcinoma (CCA) emphasize a requirement for new effective compounds for the treatment of this malignancy. This study aimed to investigate the antiproliferative property of cucurbitacin B on KKU-100 CCA cells. The determination of
Li Yang et al.
Basic & clinical pharmacology & toxicology, 127(5), 371-379 (2020-05-27)
The present study investigated the improving effect of cucurbitacin B on liver fibrosis induced by concanavalin A in mice and explored its possible mechanism. AST, ALT and TB were detected by kits. ELISA was performed to detect the levels of
S B Patel et al.
Scientific reports, 10(1), 2109-2109 (2020-02-09)
Diplocyclos palmatus (L.) C. Jeffrey is an important medicinal plant used in several reproductive medicines. It serves as a wide source of tetracyclic triterpens called cucurbitacins. Response surface methodology (RSM) with Box-Behnken design (BBD) was studied to optimize the production
Imran Khan et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 16(8), 1022-1043 (2020-02-01)
Glioblastoma is one of the most aggressive and devastating tumours of the central nervous system with short survival time. Glioblastoma usually shows fast cell proliferation and invasion of normal brain tissue causing poor prognosis. The present standard of care in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service