Skip to Content
Merck
All Photos(1)

Key Documents

P8074

Sigma-Aldrich

TWEEN® 80

BioXtra, viscous liquid

Synonym(s):

TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

description

non-ionic

Quality Level

product line

BioXtra

form

viscous liquid

mol wt

micellar avg mol wt 79,000
average mol wt 1310

composition

Oleic acid, ~70% (balance primarily linoleic, palmitic, and stearic acids)

aggregation number

60

technique(s)

DNA extraction: suitable
RNA extraction: suitable
cell culture | mammalian: suitable
protein purification: suitable

impurities

≤0.025% Phosphorus (P)

ign. residue

<0.1%

CMC

0.012 mM (20-25°C)

transition temp

cloud point 65 °C

density

1.06 g/mL at 25 °C

anion traces

chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.001%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.05%
Pb: <0.001%
Zn: <0.0005%

HLB

15

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

["TWEEN"] 80 is a polyethylene sorbitol ester, broadly used in biochemical applications like solubilizing proteins, isolating nuclei from cells in culture, and growing tubercle bacilli. It is also useful in emulsifying and dispersing substances in medicinal and food products. TWEEN 80 acts as an active drying agent in ice cream.

Application

["TWEEN"] 80 has been used:
  • to dissolve rapamycin (RAPA) for immunosuppressive treatment to perform in vivo studies in mice
  • to dissolve remodelin and remodelin Fluor to assess remodelin toxicity in vivo and small molecule dosing
  • to prepare assay medium for the microbiological assay for total folate

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey D Keeler et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(1), 173-183 (2017-09-26)
The devastating neurodegenerative disease multiple sclerosis (MS) could substantially benefit from an adeno-associated virus (AAV) immunotherapy designed to restore a robust and durable antigen-specific tolerance. However, developing a sufficiently potent and lasting immune-regulatory therapy that can intervene in ongoing disease
Joel Selkrig et al.
Scientific reports, 8(1), 10646-10646 (2018-07-15)
In animals, commensal microbes modulate various physiological functions, including behavior. While microbiota exposure is required for normal behavior in mammals, it is not known how widely this dependency is present in other animal species. We proposed the hypothesis that the
Brian Hassett et al.
mAbs, 10(1), 166-176 (2017-10-12)
Fusion protein and monoclonal antibody-based tumor necrosis factor (TNF) inhibitors represent established treatment options for a range of inflammatory diseases. Regulatory authorities have outlined the structural characterization and clinical assessments necessary to establish biosimilarity of a new biotherapeutic product with
Hidetaka Tanno et al.
Science advances, 6(17), eaay9093-eaay9093 (2020-05-20)
Natively paired sequencing (NPS) of B cell receptors [variable heavy (VH) and light (VL)] and T cell receptors (TCRb and TCRa) is essential for the understanding of adaptive immunity in health and disease. Despite many recent technical advances, determining the
Sen Zhu et al.
Nature communications, 9(1), 500-500 (2018-02-07)
BMI1, a polycomb group (PcG) protein, plays a critical role in epigenetic regulation of cell differentiation and proliferation, and cancer stem cell self-renewal. BMI1 is upregulated in multiple types of cancer, including prostate cancer. As a key component of polycomb

Articles

A complete reversed phase HPLC-ELSD workflow, including sample preparation for the analysis of Polysorbate 80 in a monoclonal antibody formulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service