Skip to Content
Merck
All Photos(2)

Key Documents

P7045

Supelco

Phenformin hydrochloride

analytical standard

Synonym(s):

N-(2-Phenylethyl)imidodicarbonimidic diamide monohydrochloride, Phenethylbiguanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5 · HCl
CAS Number:
Molecular Weight:
241.72
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.NC(=N)NC(=N)NCCc1ccccc1

InChI

1S/C10H15N5.ClH/c11-9(12)15-10(13)14-7-6-8-4-2-1-3-5-8;/h1-5H,6-7H2,(H6,11,12,13,14,15);1H

InChI key

YSUCWSWKRIOILX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenformin hydrochloride is a biguanide derivative that shows anti-diabetic activity.

Application

Phenformin hydrochloride has been used as a test compound to study the antineoplastic activity it can exhibit in suppressing the growth of ovarian cancer cells and also human hepatocellular carcinoma cells.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relevance of the OCT1 transporter to the antineoplastic effect of biguanides
Segal D E, et al.
Biochemical and Biophysical Research Communications, 414(4), 694-699 (2011)
Energy metabolism determines the sensitivity of human hepatocellular carcinoma cells to mitochondrial inhibitors and biguanide drugs
Hsu C-C, et al.
Oncology Reports, 34(3), 1620-1628 (2015)
Mozhgan Sedigh-Ardekani et al.
Regulatory toxicology and pharmacology : RTP, 70(2), 514-518 (2014-09-10)
Lactic acidosis occurs in a number of clinical conditions, e.g. in surgeries, orthotopic liver transplant, and anesthetic agent administration, which has deleterious effects on the patient's survival. The most rational therapy for these patients, the sodium bicarbonate administration, cannot prevent
Kevin R W Ngoei et al.
Cell chemical biology, 25(6), 728-737 (2018-04-17)
The AMP-activated protein kinase (AMPK) αβγ heterotrimer regulates cellular energy homeostasis with tissue-specific isoform distribution. Small-molecule activation of skeletal muscle α2β2 AMPK complexes may prove a valuable treatment strategy for type 2 diabetes and insulin resistance. Herein, we report the
Valentina R Garbarino et al.
Pharmacological research, 140, 21-32 (2018-11-14)
Poorly managed gestational diabetes can lead to severe complications for mother and child including fetal overgrowth, neonatal hypoglycemia and increased autism risk. Use of metformin to control it is relatively new and promising. Yet safety concerns regarding gestational metformin use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service