Skip to Content
Merck
All Photos(4)

Documents

P3504

Sigma-Aldrich

Ponceau S

Powder

Synonym(s):

3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
Colour Index Number:
27195
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Ponceau S, BioReagent, suitable for electrophoresis

product line

BioReagent

Quality Level

form

powder

technique(s)

electrophoresis: suitable

color

red to very dark red

solubility

H2O: 1 mg/mL

ε (extinction coefficient)

≥330000 at 518-522 nm in deionized water at 0.015 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ponceau S is the sodium salt of a diazo dye. It is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. The stain is reversed with water washes and aids in subsequent immunological detection.
The following are two common stain formulations:
0.1% Ponceau S (w/v) in 5% acetic acid
2% Ponceau S (w/v) in 30% TCA, 30% sulfosalicylic acid

Application

Ponceau S has been used for membrane staining (Western blotting.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sex difference in the expression of DNA methyltransferase 3a (DNMT3a) in the rat amygdala during development
MH Kolodkin and AP Auger
Journal of Neuroendocrinology (2011)
Characterization of stress response in human retinal epithelial cells
Journal of Cellular and Molecular Medicine (2012)
Bernard L. Horecker, Earl R. Stadtman
Current Topics in Cellular Regulation (2014)
Ionizing Radiation?dependent ?-H2AX Focus Formation Requires Ataxia Telangiectasia Mutated and Ataxia Telangiectasia Mutated and Rad3-related
Joanna D. Friesner
Molecular Biology of the Cell (2005)
Xiaofei Wang et al.
PloS one, 9(6), e99772-e99772 (2014-06-11)
The insulin signaling pathway is critical for the control of blood glucose levels. Brown adipose tissue (BAT) has also been implicated as important in glucose homeostasis. The effect of short-term cold exposure on this pathway in BAT has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service