Skip to Content
Merck
All Photos(1)

Documents

P0310000

Penicillamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-Penicillamine, 3,3-Dimethyl-D-cysteine, 3-Mercapto-D-valine, D-(−)-2-Amino-3-mercapto-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(SH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722375
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

penicillamine

manufacturer/tradename

EDQM

mp

210 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(S)[C@@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8)/t3-/m0/s1

InChI key

VVNCNSJFMMFHPL-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Penicillamine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mona S El Raziky et al.
Journal of pediatric hematology/oncology, 36(3), 173-178 (2014-03-01)
Wilson disease (WD) is an inherited disorder of copper metabolism. Hemolytic anemia in WD occurs in up to 17% of patients at some point during their illness. To screen for WD among children presenting with hemolytic anemia. Twenty cases (mean
[D-penicillamine nephropathy].
I Ohno
Ryoikibetsu shokogun shirizu, (16 Pt 1)(16 Pt 1), 523-525 (1997-01-01)
S L Klingenberg et al.
The Cochrane database of systematic reviews, (1)(1), CD004182-CD004182 (2006-01-27)
Primary sclerosing cholangitis is a cholestatic disease. D-penicillamine is suggested as a treatment option due to its copper reducing and immunomodulatory potential. To evaluate the beneficial and harmful effects of D-penicillamine for patients with primary sclerosing cholangitis. Eligible trials were
Rim Ishak et al.
American journal of clinical dermatology, 14(3), 223-233 (2013-04-23)
Penicillamine is a well-known heavy metal chelator, classically used in the treatment of Wilson disease, rheumatoid arthritis, and cystinuria. From a dermatologic standpoint, penicillamine was found to be useful in the treatment of systemic sclerosis. The successful therapeutic uses of
C U Manzini et al.
Clinical rheumatology, 31(1), 183-187 (2011-10-13)
Eosinophilic fasciitis (EF) is a rare disease characterized by symmetrical thickness and hardening of the skin, especially localized to forearms and thorax, with eosinophilia. Corticosteroids represent the first-line therapy, even if some patients are scarcely responsive and/or may develop important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service