Skip to Content
Merck
All Photos(1)

Key Documents

P0270000

Papaverine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4 · HCl
CAS Number:
Molecular Weight:
375.85
Beilstein:
3921435
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

papaverine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC1=C(OC)C=C(C(CC2=CC(OC)=C(OC)C=C2)=NC=C3)C3=C1.Cl

InChI

1S/C20H21NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-8,10-12H,9H2,1-4H3;1H

InChI key

UOTMYNBWXDUBNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Papaverine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Smooth muscle relaxant and cerebral vasodilator; phosphodiesterase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Desgagné-Penix et al.
The Plant journal : for cell and molecular biology, 72(2), 331-344 (2012-06-26)
Papaverine, a major benzylisoquinoline alkaloid in opium poppy (Papaver somniferum), is used as a vasodilator and antispasmodic. Conversion of the initial intermediate (S)-norcoclaurine to papaverine involves 3'-hydroxylation, four O-methylations and dehydrogenation. However, our understanding of papaverine biosynthesis remains controversial more
J M Mathis et al.
Neuroradiology, 39(2), 90-98 (1997-02-01)
Intra-arterial infusion of papaverine hydrochloride for subarachnoid hemorrhage-induced cerebral vasospasm has become an adjunctive endovascular therapy along with cerebral angioplasty. Our knowledge concerning the mechanism of action, method of administration and potential side effects of this therapeutic alternative are reviewed.
Seyed Alaeddin Asgari et al.
BJU international, 110(3), 449-452 (2012-02-22)
• To assess the efficacy of papaverine hydrochloride combined with a diclofenac sodium suppository to relieve renal colic compared with diclofenac suppository monotherapy, as the effect of phosphodiesterase inhibitors on ureteric muscles might reduce the pain of renal colic. •
Shaukat Nawaz Khan et al.
The Cochrane database of systematic reviews, 10, CD003635-CD003635 (2012-10-19)
During aortic aneurysm surgery, cross-clamping can lead to inadequate blood supply to the spinal cord resulting in neurological deficit. Cerebrospinal fluid drainage (CSFD) may increase the perfusion pressure to the spinal cord and hence reduce the risk of ischaemic spinal
Brian Lima et al.
The Journal of thoracic and cardiovascular surgery, 143(4), 945-952 (2012-02-22)
An array of neuroprotective strategies has evolved to limit spinal cord injury during descending thoracic aneurysm and thoracoabdominal aortic aneurysm repair. This study prospectively assessed the neuroprotective impact of intrathecal papaverine added to other techniques in aortic aneurysm repairs. From

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service