Skip to Content
Merck
All Photos(6)

Key Documents

O9882

Sigma-Aldrich

Octyl β-D-glucopyranoside

BioXtra, ≥98% (GC)

Synonym(s):

n-Octyl glucoside, OGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Beilstein:
84118
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥98% (GC)

form

powder

mol wt

micellar avg mol wt 25,000

aggregation number

84

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.2%

CMC

20-25 mM (20-25°C)

transition temp

cloud point >100 °C

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.003%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.1%
Na: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic, dialyzable detergent for the solubilization and isolation of membrane proteins. Has been shown to increase the resolution of proteins in 2D gels.
Octyl β --D-glucopyranoside has been used as a component of aqueous buffer containing NaCl, HEPES and chromogenic substrate for use in trypsin assays

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potent, nonpeptide inhibitors of human mast cell tryptase. Synthesis and biological evaluation of novel spirocyclic piperidine amide derivatives.
Bioorganic & Medicinal Chemistry Letters, 18(6), 2114-2121 (2008)
Chaoqun Yao et al.
Proteomics. Clinical applications, 4(1), 4-16 (2010-12-08)
About two million new cases of leishmaniasis with 50 000 associated deaths occur worldwide each year. Promastigotes of the causative Leishmania spp. develop from the procyclic stage to the highly virulent metacyclic stage within the sand fly vector. We hypothesized
Christine Ménager et al.
Langmuir : the ACS journal of surfaces and colloids, 26(19), 15453-15463 (2010-09-10)
The present study deals with the morphological modifications of giant dioleoyl phosphatidylcholine vesicles (DOPC GUVs) induced by the nonionic surfactant n-octyl β,D-glucopyranoside at sublytic levels, i.e., in the first steps of the vesicle-to-micelle transition process, when surfactant inserts into the
Shigesaburo Ogawa et al.
Carbohydrate research, 345(17), 2534-2541 (2010-10-05)
The lyotropic behavior and glass-forming properties of octyl β-D-glucoside (C8Glu) and octyl β-D-thioglucoside (C8SGlu)/water binary mixtures were evaluated using differential scanning calorimetry (DSC) and polarizing optical microscopy (POM). The results clearly indicate that the mixture forms a glass in the
Kerry K Karukstis et al.
The journal of physical chemistry. B, 116(12), 3816-3822 (2012-03-06)
When mixed with water, n-octyl β-D-glucoside forms self-assembled nanostructures, several of which are liquid crystalline and all of which depend on the water/glucoside ratio and temperature. For practical use of these phases, a detailed understanding of the conditions under which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service