Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8398

Human DNA for whole-genome variant assessment (Daughter of Utah/European Ancestry)

NIST SRM 8398

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

manufacturer/tradename

NIST®

application(s)

genomic analysis

General description

This reference material consists of a female whole human genome sample of Utah/European ancestry. A unit of RM 8398 consists of a vial containing human genomic DNA extracted from a single large growth of human lymphoblastoid cell line GM12878 (labeled as HG-001). The vial contains approximately 10μg of genomic DNA, and the DNA is in TE buffer (10 mM TRIS, 1 mM EDTA, pH 8.0).

SRM 8398_cert

SRM 8398_SDS

Application

Standard Reference Material (SRM) intended for validation, optimization, and process evaluation purposes. Used to assess performance of human genome sequencing variant calling by obtaining estimates of true positives, false positives, and false negatives.
Sequencing applications could include whole genome sequencing, whole exome sequencing, and more targeted sequencing such as gene panels. This genomic DNA is intended to be analyzed in the same way as any other sample a lab would process and analyze extracted DNA.

Features and Benefits

  • RM 8398 is from a human lymphoblastoid cell line and is intended for research use.
  • Details of expiration date, stability, homogeneity, and storage and use instructions are provided on NIST certificate.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service