Skip to Content
Merck
All Photos(2)

Key Documents

NIST2899A

Ethanol solution

NIST® SRM® 2899a, nominal 25% by mass

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

concentration

(nominal mass fraction 25%)

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 2899A contains five 10-milliliter ampoules, each consisting of ~10 mL of solution. For more information, please refer to the SDS and COA.

SRM 2899A_cert

SRM 2899A_SDS

Application

Standard reference material Ethanol solution is primarilyintended for calibrating instruments and techniques used in the determinationof ethanol in breath.

Features and Benefits

  • Standard reference material Ethanol solution is primarily intended for calibrating instruments and techniques used in the determination of ethanol in breath.
  • The NIST-certified mass fraction values of ethanol are traceable to the SI derived unit of mass fraction and are expressed as a percentage.
  • Expiration Details and instructions for handling, storage and use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Ethanol

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Ding et al.
Nanomedicine : nanotechnology, biology, and medicine, 13(1), 297-305 (2016-10-05)
Au@Ag nanoparticles (NPs) were recently found to display giant two-photon photoluminescence (2PPL) enhancement, with an enhancement factor up to 815 fold upon aggregate formation. Based on this finding, two-photon imaging of bacteria by Au@Ag NPs under near-infrared (NIR) femtosecond laser
Fatma Trabelsi et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 69(6), 349-358 (2017-03-07)
This study investigates the cytotoxicity and the genotoxicity induced by arsenic trioxide As
Tomomi Sanagi et al.
Frontiers in cellular neuroscience, 13, 344-344 (2019-08-17)
Autism spectrum disorder (ASD) is one of the most widespread neurodevelopmental disorders, characterized by impairment in social interactions, and restricted stereotyped behaviors. Using immunohistochemistry and positron emission tomography (PET), several studies have provided evidence of the existence of activated microglia
Jinhui Li et al.
Materials (Basel, Switzerland), 11(5) (2018-05-05)
Thin BaTiO₃ (BT) coating layers are required in various multilayer ceramic technologies, and fine nanosized BT particles with good dispersion in solution are essential for this coating process. In this work, cubic and tetragonal phase monodispersed BT nanoparticles—which were referred
Hongdan Ma et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-19)
Paeonol exhibits a wide range of pharmacological activities, such as anti-inflammatory, antidiabetic as well as pain-relieving activities. However, its intrinsic properties, such as low water solubility, poor stability and low oral bioavailability, restrict its clinical application. The current study aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service