Skip to Content
Merck
All Photos(1)

Key Documents

N0750000

Nifedipine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O6
CAS Number:
Molecular Weight:
346.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nifedipine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2ccccc2[N+]([O-])=O)C(=O)OC

InChI

1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3

InChI key

HYIMSNHJOBLJNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nifedipine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neeraj Dhaun et al.
Journal of the American Society of Nephrology : JASN, 24(1), 31-36 (2012-12-18)
Arterial stiffness and impaired nitric oxide (NO) bioavailability contribute to the high risk for cardiovascular disease in CKD. Both asymmetric dimethylarginine (ADMA), an endogenous inhibitor of NO production, and endothelin-1 (ET-1) oppose the actions of NO, suggesting that ET-1 receptor
Joel S Klinck et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 157(2), 236-250 (2012-12-04)
Transport of calcium (Ca) and cadmium (Cd) was examined along the gastro-intestinal tract (GIT) of freshwater and seawater Oncorhynchus mykiss irideus (FWT and SWTies respectively) using in vitro and in vivo experiments. Based on known physiological differences between FWT and
Carolien Roos et al.
JAMA, 309(1), 41-47 (2013-01-03)
In threatened preterm labor, maintenance tocolysis with nifedipine, after an initial course of tocolysis and corticosteroids for 48 hours, may improve perinatal outcome. To determine whether maintenance tocolysis with nifedipine will reduce adverse perinatal outcomes due to premature birth. APOSTEL-II
Yui Yamamoto et al.
Journal of pharmacological sciences, 121(3), 212-226 (2013-03-02)
We recently developed a novel cognitive enhancer, ST101 (spiro[imidazo[1,2-a]pyridine-3,2-indan]-2(3H)-one), that activates T-type voltage-gated calcium channels (VGCCs). Here, we address whether T-type VGCC activation with ST101 mediates its cognitive effects in vivo and the relevance of T-type VGCC activation to acetylcholine
Corey B Toal et al.
Blood pressure, 21 Suppl 1, 3-10 (2012-07-06)
Calcium-channel blockers (CCBs) constitute a diverse group of compounds but are often referred to as a single homogeneous class of drug and the clinical responses indiscriminately summarized. Even within the dihydropyridine subgroup, there are significant differences in formulations, pharmacokinetics, durations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service