Skip to Content
Merck
All Photos(1)

Documents

N0300000

Neamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Deoxy-4-O-(2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl)-D-streptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26N4O6
CAS Number:
Molecular Weight:
322.36
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

neomycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C12H26N4O6/c13-2-5-8(18)9(19)6(16)12(21-5)22-11-4(15)1-3(14)7(17)10(11)20/h3-12,17-20H,1-2,13-16H2/t3-,4+,5-,6-,7+,8-,9-,10-,11-,12-/m1/s1

InChI key

SYJXFKPQNSDJLI-HKEUSBCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia. Their suitability for any other use is not guaranteed and is the sole responsibility of the user. This standard is not intended for human or animal use.
Established for the preparation of reference solutions (d) in the testing of related substances in framycetin sulfate and neomycin sulfate using liquid chromatography (General text 2.2.29), according to the monographs 0180 and 0197 of European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alok Upadhyay et al.
Molecular & cellular proteomics : MCP, 12(6), 1539-1552 (2013-02-23)
Hepatitis C virus (HCV) infection leading to chronic hepatitis is a major factor in the causation of liver cirrhosis, hepatocellular carcinoma, and liver failure. This process may involve the interplay of various host cell factors, as well as the interaction
Isabelle Baussanne et al.
Journal of medicinal chemistry, 53(1), 119-127 (2009-12-17)
The aminoglycoside antibiotics bind to the 16S bacterial rRNA and disturb the protein synthesis. One to four hydroxyl functions of the small aminoglycoside neamine were capped with phenyl, naphthyl, pyridyl, or quinolyl rings. The 3',4'- (6), 3',6- (7a), and the
Regioselective diazo-transfer reaction at the C3-position of the 2-desoxystreptamine ring of neamine antibiotics.
Andreas A Bastian et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(28), 9151-9154 (2013-05-29)
Paula López-Senín et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(6), 1946-1953 (2011-01-29)
We describe the use of dynamic combinatorial chemistry (DCC) to identify ligands for the stem-loop structure located at the exon 10-5'-intron junction of Tau pre-mRNA, which is involved in the onset of several tauopathies including frontotemporal dementia with Parkinsonism linked
Tony Le Gall et al.
Bioconjugate chemistry, 20(11), 2032-2046 (2009-10-24)
With the view to develop novel bioinspired nonviral vectors for gene delivery, we synthesized a series of cationic lipids with a neamine headgroup, which incorporates rings I and II of the natural antibiotic aminoglycoside neomycin B. Indeed, we reasoned that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service