Skip to Content
Merck
All Photos(4)

Key Documents

M9647

Sigma-Aldrich

D-Mannitol

ACS reagent

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

powder

optical activity

[α]25/D +23.3 to +24.3°(lit.)

impurities

Reducing sugars, passes test
≤0.0008 meq/g Titratable acid
≤0.01% Insoluble matter

ign. residue

≤0.01%

loss

≤0.05% loss on drying, 105°C

color

white

useful pH range

5.0-6.5 (25 °C, 182 g/L)

mp

167-170 °C (lit.)

solubility

water: soluble 182 g/L at 20 °C (68 °F)

cation traces

heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ping Wang et al.
Autophagy, 16(1), 123-139 (2019-03-27)
Autophagy is a conserved catabolic process in eukaryotes that contributes to cell survival in response to multiple stresses and is important for organism fitness. In Arabidopsis thaliana, the core machinery of autophagy is well defined, but its transcriptional regulation is
Diana Bilton et al.
Thorax, 69(12), 1073-1079 (2014-09-24)
Bronchiectasis is characterised by excessive production of mucus and pulmonary exacerbations. Inhaled osmotic agents may enhance mucociliary clearance, but few long-term clinical trials have been conducted. To determine the impact of inhaled mannitol on exacerbation rates in patients with non-cystic
Heikki O Koskela et al.
Chest, 124(4), 1341-1349 (2003-10-14)
To compare the sensitivity and validity of mannitol, histamine, and cold air challenges to demonstrate the effect of inhaled corticosteroids (ICS) in asthma. A prospective study. Seventeen patients with recently diagnosed, steroid-naive asthma who fulfilled the diagnostic criteria of Finnish
Heikki O Koskela et al.
Chest, 125(6), 1985-1992 (2004-06-11)
To define whether coughing during mannitol challenge is a nonspecific side effect of this challenge or is associated with asthma. A prospective study. University hospital. Thirty-seven steroid-naive, asthmatic subjects and 10 healthy subjects. The participants completed a symptom questionnaire, recorded
Cecilio F Caldeira et al.
Nature communications, 5, 5365-5365 (2014-11-06)
Circadian rhythms enable plants to anticipate daily environmental variations, resulting in growth oscillations under continuous light. Because plants daily transpire up to 200% of their water content, their water status oscillates from favourable during the night to unfavourable during the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service