Skip to Content
Merck
All Photos(1)

Key Documents

M1300000

Methylatropine bromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Atropine methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26NO3 · Br
CAS Number:
Molecular Weight:
384.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atropine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Br.C[N]1(C)[C@@H]2CC[C@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C18H26NO3.BrH/c1-19(2)14-8-9-15(19)11-16(10-14)22-18(21)17(12-20)13-6-4-3-5-7-13;/h3-7,14-17,20H,8-12H2,1-2H3;1H/t14-,15+,16?,17?;

InChI key

YBFLMRSYEWDJEJ-ZNHDNBJUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylatropine bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Siew Kian Tai et al.
Hippocampus, 22(4), 914-925 (2011-05-05)
The vestibular system has been suggested to participate in spatial navigation, a function ascribed to the hippocampus. Vestibular stimulation during spatial navigation activates a hippocampal theta rhythm (4-10 Hz), which may enhance spatial processing and motor response. We hypothesize that
Bo Ahrén et al.
Peptides, 125, 170152-170152 (2019-09-17)
Glucose-dependent insulinotropic polypeptide (GIP) receptor knockout (KO) mice are tools for studying GIP physiology. Previous results have demonstrated that these mice have impaired insulin response to oral glucose. In this study, we examined the insulin response to intravenous glucose by
Bashair M Mussa et al.
Brain research, 1394, 62-70 (2011-05-03)
We and others have previously shown that the dorsal motor nucleus of the vagus (DMV) is involved in regulation of pancreatic exocrine secretion. Many pancreatic preganglionic neurons within the DMV are inhibited by pancreatic secretagogues suggesting that an inhibitory pathway
Takahiko Yoshimoto et al.
The journal of physiological sciences : JPS, 61(5), 373-383 (2011-06-30)
In this study, after confirming the suppression of autonomic nervous function by isoflurane anesthesia using autonomic antagonists, we pharmacologically investigated the involvement of vasomotor and cardiac sympathetic functions in systolic blood pressure variability (SBPV) frequency components in conscious rats at
Apurva Kumar R Joshi et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 64(1-2), 115-120 (2010-08-03)
The purpose of this study was to investigate the involvement of acetylcholinesterase (AChE) inhibition in hyperglycemic and stressogenic effects of monocrotophos in rats. Oral administration of monocrotophos (1.8 mg/kg b.w., 1/10 LD(50)) caused reversible hyperglycemia in rats with peak increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service