Skip to Content
Merck
All Photos(1)

Documents

M0370020

Mepivacaine impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Desbutylbupivacaine, (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mepivacaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(NC1=C(C)C=CC=C1C)C2NCCCC2

InChI

1S/C14H20N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17)

InChI key

SILRCGDPZGQJOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mepivacaine impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gantenbein et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(4), 383-385 (2000-03-22)
Bupivacaine is used to provide prolonged anesthesia and postoperative analgesia. The human cytochrome P450 (CYP) involved in bupivacaine degradation into pipecolylxylidine (PPX), its major metabolite, has, to our knowledge, never been described. Microsome samples were prepared from six human livers
S Reif et al.
Journal of chromatography. B, Biomedical sciences and applications, 719(1-2), 239-244 (1998-12-30)
A sensitive HPLC method has been developed for the determination of ropivacaine, 3-hydroxy-ropivacaine, 4-hydroxy-ropivacaine and 2',6'-pipecoloxylidide in plasma. The procedure involved extraction from plasma with a mixture of n-heptane-ethyl acetate and a back-extraction into an acidified aqueous solution. The chromatography
J Butterworth et al.
Anesthesia and analgesia, 85(2), 336-342 (1997-08-01)
Local anesthetics inhibit binding of ligands to beta2-adrenergic receptors (beta2ARs), and, as a consequence, inhibit intracellular cAMP production. We hypothesized that among homologous local anesthetics, their avidity at inhibiting binding of tritiated dihydroalprenolol (3H-DHA) to beta2ARs would increase with increasing
P J Pere et al.
British journal of anaesthesia, 106(4), 512-521 (2011-02-11)
As ropivacaine and its metabolites are excreted by the kidneys, we studied their disposition in subjects with renal dysfunction. Twenty patients with moderate or severe renal insufficiency and 10 healthy volunteers received ropivacaine 1 mg kg(-1) i.v. over 30 min.
T Arvidsson et al.
Chirality, 7(4), 272-277 (1995-01-01)
Ropivacaine hydrochloride monohydrate (ropivacaine) is a new local anaesthetic agent which is administered exclusively as the (-)-(S)-form. The aim of the study was to determine whether metabolic racemisation of (-)-(S)-ropivacaine occurs. This was tested in man, rat, dog, and sheep

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service