Skip to Content
Merck
All Photos(1)

Key Documents

IRMM522C

Copper

IRMM®, certified reference material, 0.5 mm wire

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

resistivity

1.673 μΩ-cm, 20°C

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Analysis Note

For more information please see:
IRMM522C

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kessarin Thanapirom et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96(2), 150-156 (2013-08-14)
Wilson's disease (WD) is an inherited disorder of copper metabolism, leading to hepatic and neuropsychiatric manifestations. The long-term outcome of patients with Wilson's disease presenting with acute liver failure (ALF) versus those with acute-on-chronic liver failure (ACLF) has not been
Marius Andruh
Chimia, 67(6), 383-387 (2013-08-16)
The node-and-spacer approach is widely employed in crystal engineering to construct coordination polymers. It consists of self-assembly processes involving mononuclear cationic species and exo-dentate ligands. We enlarged this strategy using preformed homo- and heterometallic complexes as nodes. The presence of
Xuejiao Tong et al.
Journal of environmental sciences (China), 25(4), 652-658 (2013-08-09)
The removal efficiency of copper (Cu(II)) from an actual acidic electroplating effluent by biochars generated from canola, rice, soybean and peanut straws was investigated. The biochars simultaneously removed Cu(II) from the effluent, mainly through the mechanisms of adsorption and precipitation
Ping Ning et al.
Journal of environmental sciences (China), 25(4), 808-814 (2013-08-09)
Metal (Cu, Co, or Zn) loaded ZSM-5 and Y zeolite adsorbents were prepared for the adsorption of hydrogen cyanide (HCN) toxic gas. The results showed that the HCN breakthrough capacity was enhanced significantly when zeolites were loaded with Cu. The
Yaser A Almaroai et al.
Environmental technology, 34(5-8), 747-755 (2013-07-11)
Kinetic aspects of metal release were investigated to understand the effects of synthetic chelating agents (EDTA, EDDS and NTA) and low molecular weight (LMW) organic acids (oxalic and citric acids) on the release kinetics of Cr, Cu and As in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service