Skip to Content
Merck
All Photos(1)

Key Documents

I0500000

Isoniazid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Pyridinecarboxylic acid hydrazide, INH, Isonicotinic acid hydrazide, Isonicotinic hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
Beilstein:
119374
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

isoniazid

manufacturer/tradename

EDQM

mp

171-173 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NNC(=O)c1ccncc1

InChI

1S/C6H7N3O/c7-9-6(10)5-1-3-8-4-2-5/h1-4H,7H2,(H,9,10)

InChI key

QRXWMOHMRWLFEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoniazid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Antibiotic for treatment of Mycobacterium tuberculosis, inhibits mycolic acid biosynthesis. Metabolized by hepatic N-acetyltransferase (NAT) and cytochrome P450 2E1 (CYP2E1) to form hepatotoxins. Selectively induces expression of CYP2E1. Reversibly inhibits CYP2C19 and CYP3A4 activities, and mechanistically inactivates CYP1A2, CYP2A6, CYP2C19 and CYP3A4 at clinically relevant concentrations.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laura Muñoz et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 60(3), 349-356 (2014-10-15)
The extent to which anti-tumor necrosis factor (TNF)-associated tuberculosis can be prevented is unclear, and there is no established guidance on the optimal screening strategy for latent tuberculosis (LTBI) in patients about to start anti-TNF therapy. We aimed to determine
Anandi Martin et al.
The Journal of antimicrobial chemotherapy, 62(1), 56-64 (2008-04-15)
The reference standard methods for drug susceptibility testing (DST) of M. tuberculosis are very slow to give results, and due to the emergence of multidrug-resistant tuberculosis (MDR-TB) and extensively drug-resistant tuberculosis, there is an urgent demand for new, rapid and
Molebogeng X Rangaka et al.
Lancet (London, England), 384(9944), 682-690 (2014-05-20)
Antiretroviral therapy reduces the risk of tuberculosis, but tuberculosis is more common in people with HIV than in people without HIV. We aimed to assess the effect of isoniazid preventive therapy on the risk of tuberculosis in people infected with
A fresh look at the mechanism of isoniazid-induced hepatotoxicity.
I G Metushi et al.
Clinical pharmacology and therapeutics, 89(6), 911-914 (2011-03-18)
Andrew D Kerkhoff et al.
PloS one, 7(11), e49928-e49928 (2012-12-05)
People living with HIV (PLWH) who have positive tuberculin skin tests (TST) benefit from isoniazid preventive therapy (IPT) whereas those testing TST-negative do not. Revised World Health Organization guidelines explicitly state that assessment of TST is not a requirement for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service