Skip to Content
Merck
All Photos(1)

Key Documents

I0086000

Imidazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor pressure

<1 mmHg ( 20 °C)

API family

imidazole

manufacturer/tradename

EDQM

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Excellent for buffers in the range of pH 6.2-7.8

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleonora Petryayeva et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 977-987 (2013-01-10)
Methods have been developed for the solid-phase detection of nucleic acids using mixed films of quantum dots (QDs) and oligonucleotide probes in microtiter plates. Polystyrene microwells were functionalized with multidentate imidazole ligands to immobilize QDs. Oligonucleotide hybridization was transduced using
Joy Rathjen et al.
Reproduction, fertility, and development, 26(5), 703-716 (2013-06-14)
Human embryonic stem (ES) cells have been proposed as a renewable source of pluripotent cells that can be differentiated into various cell types for use in research, drug discovery and in the emerging area of regenerative medicine. Exploitation of this
Aviva Levina et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(11), 3609-3619 (2013-01-31)
An anti-metastatic drug, NAMI-A ((ImH)[Ru(III) Cl4 (Im)(dmso)]; Im=imidazole, dmso=S-bound dimethylsulfoxide), and a cytotoxic drug, KP1019 ((IndH)[Ru(III) Cl4 (Ind)2 ]; Ind=indazole), are two Ru-based anticancer drugs in human clinical trials. Their reactivities under biologically relevant conditions, including aqueous buffers, protein solutions
Gabriel T M Mashabela et al.
Chemical communications (Cambridge, England), 49(70), 7714-7716 (2013-07-24)
OvoA is an iron(II) dependent sulfoxide synthase which catalyzes the first step in ovothiol A biosynthesis. This enzyme sulphurizes the C5 position of the imidazole side chain of L-histidine. We report the substrate specificity profile of this catalyst and present
Ching-Wen Lin et al.
Biomaterials, 34(17), 4387-4393 (2013-03-13)
Chemically modified antisense RNA oligonucleotides (antagomir) offer promise for cancer therapies but suffer from poor therapeutic effect after systemic administration. Chemical modification or loading in degradable hydrogels can offer improvements in the accuracy and efficacy for sustained delivery at specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service