Skip to Content
Merck
All Photos(2)

Key Documents

ED2P

Sigma-Aldrich

Ethylenediaminetetraacetic acid dipotassium salt dihydrate

≥98%

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid dipotassium salt, Dipotassium ethylenediaminetetraacetate dihydrate, EDTA dipotassium salt, Edathamil, Potassium ethylenediaminetetraacetate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2(KOOCCH2)NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
404.45
EC Number:
UNSPSC Code:
12352107
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98%

form

powder

reaction suitability

reagent type: chelator

pH

4.0-5.0 (25 °C, 40.4 g/L)

mp

255-280 °C (dec.) (lit.)

solubility

3 M KOH: 160 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

O.O.O.O.[K+].[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

JFROQFOCFOKDKU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ethylenediaminetetraacetic acid dipotassium salt dehydrate has been used as a component:
  • of EDTA–glycerol solution (EDTA-G) for decalcification
  • of buffer for monoamine and pterine analysis
  • of T-cell resuspension buffer

Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Biochem/physiol Actions

Ethylenediaminetetraacetic acid (EDTA) is an anticoagulant, majorly used in preventing clotting of blood. EDTA may trigger aggregation of platelets resulting in pseudothrombocytopenia. EDTA-tetracycline therapy decreases coronary artery calcium levels in atherosclerosis. Calcium disodium EDTA administration is effective in slowing down the progression of chronic kidney disease. It also enhances the antimicrobial functionality of lysozyme and improves shelf life of cheese.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whole body insulin responsiveness is higher in beef steers selected for increased muscling
McGilchrist P, et al.
Animal : An International Journal of Animal Bioscience, 5(10), 1579-1586 (2011)
A novel decalcification method for adult rodent bone for histological analysis of peripheral-central nervous system connections
Begum F, et al.
Journal of Neuroscience Methods, 187(1), 59-66 (2010)
Cytochrome c oxidase activity is a metabolic checkpoint that regulates cell fate decisions during T cell activation and differentiation
Tarasenko TN, et al.
Cell Metabolism, 25(6), 1254-1268 (2017)
Comparison of the efficacy of ethylenediaminetetraacetic acid and tetracycline hydrochloride as root conditioning agents: An in vitro study
Nanda T, et al.
Journal of the International Clinical Dental Research Organization, 4(1), 2-2 (2012)
Lysozyme/EDTA disodium salt and modified-atmosphere packaging to prolong the shelf life of burrata cheese
Conte A, et al.
Journal of Dairy Science, 94(11), 5289-5297 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service