Skip to Content
Merck
All Photos(1)

Documents

E1515000

Erythropoietin

BRP, European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107

API family

erythropoietin

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Erythropoietin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choice of erythropoiesis stimulating agent in ESRD.
Steven Fishbane et al.
Nephrology news & issues, 27(7), Supp 10-Supp 12 (2013-07-17)
Peginesatide for anemia in chronic kidney disease.
Haruo Tomoda
The New England journal of medicine, 368(16), 1553-1553 (2013-04-19)
Management of anemia in patients with kidney disease in 2013 and beyond.
Jay B Wish
Nephrology news & issues, 27(7), Supp 1-Supp 3 (2013-07-17)
Is eliminating the floor for the Hb range consistent with patient-centered care?
Jeffrey S Berns
Nephrology news & issues, 27(7), Supp 15-Supp 16 (2013-07-17)
Darbepoetin alfa in systolic heart failure.
Mahir Karakas et al.
The New England journal of medicine, 369(5), 487-487 (2013-08-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service