Skip to Content
Merck
All Photos(3)

Key Documents

E1271

Sigma-Aldrich

N-Ethylmaleimide

BioXtra, ≥98% (HPLC)

Synonym(s):

NEM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
Beilstein:
112448
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Assay

≥98% (HPLC)

impurities

≤0.001% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

bp

210 °C (lit.)

mp

43-46 °C (lit.)

solubility

95% ethanol: 50 mg/mL, clear to slightly hazy (colorless to faint yellow)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

CCN1C(=O)C=CC1=O

InChI

1S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4H,2H2,1H3

InChI key

HDFGOPSGAURCEO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the covalent modification of cysteine residues in proteins.

Biochem/physiol Actions

Augments currents from native M-channels in sympathetic neurons and acts as an opener for KCNQ2, KCNQ4 and KCNQ5 channels.
Sulfhydryl alkylating agent that inactivates NADP-dependent isocitrate dehydrogenase and many endonucleases.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

163.2 °F - closed cup

Flash Point(C)

72.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuki Kurokawa et al.
Biochimica et biophysica acta, 1838(1 Pt B), 382-387 (2013-10-22)
Hv1 (also named, voltage-sensor only protein, VSOP) lacks an authentic pore domain, and its voltage sensor domain plays both roles in voltage sensing and proton permeation. The activities of a proton channel are intrinsic to protomers of Hv1, while Hv1
Nicolas Matuszak et al.
Journal of medicinal chemistry, 52(23), 7410-7420 (2009-07-09)
The endocannabinoid 2-arachidonoylglycerol (2-AG) plays a major role in many physiological processes, and its action is quickly terminated via enzymatic hydrolysis catalyzed by monoglyceride lipase (MGL). Regulating its endogenous level could offer therapeutic opportunities; however, few selective MGL inhibitors have
Hannah E Boycott et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(41), E3955-E3964 (2013-09-26)
Atrial myocytes are continuously exposed to mechanical forces including shear stress. However, in atrial myocytes, the effects of shear stress are poorly understood, particularly with respect to its effect on ion channel function. Here, we report that shear stress activated
Julia Kuligowski et al.
Talanta, 127, 181-190 (2014-06-11)
Liquid chromatography-mass spectrometry (LC-MS) has been increasingly used in biomedicine to study the dynamic metabolomic responses of biological systems under different physiological or pathological conditions. To obtain an integrated snapshot of the system, metabolomic methods in biomedicine typically analyze biofluids
Con Dogovski et al.
PLoS biology, 13(4), e1002132-e1002132 (2015-04-23)
Successful control of falciparum malaria depends greatly on treatment with artemisinin combination therapies. Thus, reports that resistance to artemisinins (ARTs) has emerged, and that the prevalence of this resistance is increasing, are alarming. ART resistance has recently been linked to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service