Skip to Content
Merck
All Photos(1)

Documents

D3100000

Droperidol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[1-[3-(p-Fluorobenzoyl)propyl]-1,2,3,6-tetrahydro-4-pyridyl]-2-benzimidazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22FN3O2
CAS Number:
Molecular Weight:
379.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

droperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC(=CC2)N3C(=O)Nc4ccccc34

InChI

1S/C22H22FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-11H,3,6,12-15H2,(H,24,28)

InChI key

RMEDXOLNCUSCGS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Droperidol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Min Kuo et al.
Journal of the Chinese Medical Association : JCMA, 75(5), 227-233 (2012-05-29)
Droperidol is commonly added to intravenous patient-controlled analgesia (IVPCA) regimens as an antiemetic agent. Although some studies have demonstrated its safety and efficacy, it is not clear whether adding droperidol to IVPCA infusate without an extra loading dose can effectively
G Ormel et al.
Acta anaesthesiologica Scandinavica, 55(10), 1196-1205 (2011-11-19)
Prophylactic dexamethasone, ondansetron and droperidol have a documented effect on post-operative nausea and vomiting (PONV). Still, there is a lack of studies investigating the effect of adding dexamethasone to ondansetron and droperidol in order to treat established PONV. In this
Gregory A Nuttall et al.
Anesthesiology, 118(2), 382-386 (2013-01-08)
The Food and Drug Administration issued a black box warning regarding the use of droperidol and the potential for torsade de pointes. The primary objective of this retrospective study was to determine if low-dose (0.625 mg) droperidol administration was associated
Astrid Gjelstad et al.
Analytica chimica acta, 742, 10-16 (2012-08-14)
In this paper, extraction kinetics was investigated experimentally and theoretically in hollow fiber liquid-phase microextraction (HF-LPME) and electromembrane extraction (EME) with the basic drugs droperidol, haloperidol, nortriptyline, clomipramine, and clemastine as model analytes. In HF-LPME, the analytes were extracted by
Andrey B Petrenko et al.
European journal of pharmacology, 698(1-3), 200-205 (2012-11-06)
N-methyl-d-aspartate (NMDA) receptors can be inhibited by inhalational anesthetics in vitro at clinically relevant concentrations. Here, to clarify the role of NMDA receptors in anesthetic-induced unconsciousness, we examined the hypnotic properties of isoflurane, sevoflurane and nitrous oxide in NMDA receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service