Skip to Content
Merck
All Photos(1)

Key Documents

D2963000

Doxapram hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Doxapram hydrochloride monohydrate, 1-Ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-2-pyrrolidinone monohydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30N2O2 · HCl · H2O
CAS Number:
Molecular Weight:
432.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

doxapram

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[N+H]4(CCOCC4)CCC1CN(C(=O)C1(c3ccccc3)c2ccccc2)CC.O

InChI

1S/C24H30N2O2.ClH.H2O/c1-2-26-19-22(13-14-25-15-17-28-18-16-25)24(23(26)27,20-9-5-3-6-10-20)21-11-7-4-8-12-21;;/h3-12,22H,2,13-19H2,1H3;1H;1H2

InChI key

ZOMBFZRWMLIDPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Doxapram hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Bleul et al.
Theriogenology, 73(5), 612-619 (2009-12-22)
Respiratory stimulants are widely used in asphyxic neonatal calves despite a lack of data about their effectiveness and indications of possible side effects. The effect of doxapram and theophylline on respiratory, cardiovascular, and acid-base variables was investigated in 10 healthy
A Greenough et al.
Archives of disease in childhood. Fetal and neonatal edition, 93(3), F242-F245 (2007-11-17)
Randomised trials have demonstrated that ventilation techniques which support every spontaneous breath are the most efficacious weaning modes. Nasal continuous positive airway pressure after extubation reduces the likelihood of incidents leading to the need for reintubation in very low birthweight
Guo-cai Li et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 30(2), 245-248 (2010-02-18)
To investigate the effects of doxapram on the respiratory rhythmical discharge activity (RRDA) in the brainstem slices of neonatal rats. Thirty neonatal SD rats (of either sex, 0-3 days old) were randomly divided into 6 equal groups (groups I-VI), and
Doxapram revisited in patients with cancer.
Eric Li et al.
Journal of palliative medicine, 12(8), 673-673 (2009-08-12)
Advantages of doxapram for post-anaesthesia recovery and outcomes in bariatric surgery patients with obstructive sleep apnoea.
Olumuyiwa A Bamgbade
European journal of anaesthesiology, 28(5), 387-388 (2010-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service