Skip to Content
Merck
All Photos(1)

Key Documents

C9386

Millipore

Casein Acid Hydrolysate for Biotechnology and Microbiology

for Biotechnology and Microbiology, ≥12.5% Total N, 8-12% Amino

Synonym(s):

Casein hydrolyzates, Hy-Case® SF, Peptone from casein, casein acid hydrolysate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

Product Name

Hy-Case® SF, Casein acid hydrolysate, from bovine milk

biological source

bovine milk

Quality Level

form

powder

nitrogen analysis

8-12% amino, ≥12.5% total

loss

≤11% loss on drying

pH

5.4-7.4

application(s)

food and beverages
microbiology

storage temp.

10-30°C

suitability

Streptococcus spp.
Streptomyces spp.

Looking for similar products? Visit Product Comparison Guide

General description

Hy-Case® SF is a high-quality acid hydrolysis of casein. Casein is a rich source of amino nitrogen so on treatment with a strong acid, it gets hydrolysed, producing free amino acids and several short peptide fragments. Special processing keeps inorganic ions to a minimum, so Hy-Case® SF is virtually a high concentration of free amino acids with a minimum of salt. It is a refined grade hydrolysate with high clarity in solution and is food grade and is recommended where a mixture of highly soluble free amino acids is required. It is especially recommended when inorganic ion levels need to be kept at a minimum.

Application

Casein hydrolysate Hy-Case® SF was used in DMCY medium, during an experimental study conducted to investigate the role of ssgA gene product in promoting the sporulation and cell division in Streptomyces griseus. It may also be used in the YH medium in studies related to Halobacterium denitrificans sp. which is characterised as being carbohydrate-utilizing, denitrifying, extremely halophilic bacteria.
Recommended for use as a microbiological nutrient in laboratory media, fermentations, and tissue culture applications requiring a mixture of highly soluble free amino acids.

Linkage

Similar to Hy-Case Amino, but contains a maximum of 2.0% sodium chloride

Reconstitution

Recommended concentration: 25 g/L

Legal Information

Hy-Case is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Tomlinson et al.
International journal of systematic bacteriology, 36(1), 66-70 (1986-01-01)
Halobacterium denitrificans was one of several carbohydrate-utilizing, denitrifying, extremely halophilic bacteria isolated by anaerobic enrichment in the presence of nitrate. Anaerobic growth took place only when nitrate (or nitrite) was present and was accompanied by the production of dinitrogen.
S Kawamoto et al.
Microbiology (Reading, England), 143 ( Pt 4), 1077-1086 (1997-04-01)
The ssgA gene of Streptomyces griseus B2682, when present in high copy number, results in both suppression of sporulation and fragmented growth of mycelia. Western analysis with polyclonal antibodies against the gene product (SsgA) revealed a close correlation between SsgA

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service