Skip to Content
Merck
All Photos(1)

Key Documents

C2970000

Crotamiton

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Ethyl-o-crotonotoluidide, N-Ethyl-N-(2-methylphenyl)-2-butenamide, Crotamiton

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCON(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
203.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

description

predominantly trans

API family

crotamiton

manufacturer/tradename

EDQM

refractive index

n20/D 1.54 (lit.)

bp

153-155 °C/13 mmHg (lit.)

density

0.987 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCN(C(=O)\C=C\C)c1ccccc1C

InChI

1S/C13H17NO/c1-4-8-13(15)14(5-2)12-10-7-6-9-11(12)3/h4,6-10H,5H2,1-3H3/b8-4+

InChI key

DNTGGZPQPQTDQF-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Crotamiton EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Forton et al.
The British journal of dermatology, 138(3), 461-466 (1998-05-15)
A standardized skin surface biopsy was performed in 34 patients suffering from skin diseases with high Demodex folliculorum density (Dd) > 5D/cm2 before, during and after topical treatment. The patients were randomized into six comparable groups to study six topical
Ahmed M Al-Arfaj et al.
The American journal of tropical medicine and hygiene, 76(5), 967-971 (2007-05-10)
A 31-year-old Saudi man was seen at an ear, nose, and throat clinic at Riyadh, Saudi Arabia, with bilateral itching in the external auditory canal. On otoscopic examination, the skin lining the ear canal was thickened with whitish sheets of
Habib Ansarin et al.
Dermatology online journal, 12(1), 19-19 (2006-04-28)
A wide range of clinical manifestations may be seen in scabies, from classic pruritic papules and burrows to secondary features such as impetigo. Bullus lesions are a less frequent. Twenty cases of scabies presenting with bullae have been reported so
Demodecidosis manifested on the external genitalia.
S M Hwang et al.
International journal of dermatology, 37(8), 634-636 (1998-09-10)
N Nakada et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 61(7), 1739-1747 (2010-04-08)
The behavior of antibacterial triclosan, insect-repellent diethyltoluamide (DEET), anticonvulsant carbamazepine, and antipruritic crotamiton was investigated at two sewage treatment plants (STPs) to clarify their complete mass balance. Twenty-four-hour flow-proportional composite samples were collected from the influent and effluent of primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service