Skip to Content
Merck
All Photos(3)

Documents

C0706

Sigma-Aldrich

Citric acid monohydrate

BioXtra, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.5%

form

powder

impurities

≤0.002% Phosphorus (P)
≤0.005% Insoluble matter

ash

≤0.02%

pH

1.85 (25 °C, 50 g/L)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citric acid is one of the fruits acids. It is present in various citrus fruits and vegetables.

Application

Citric acid monohydrate has been used:
  • as carbon source of carbon nanodot synthesis
  • in the preparation of citric acid monohydrate solution/buffer of pH-6.0 for tissue sample preparation
  • to treat various samples in citric acid solubilisation technique

Biochem/physiol Actions

Citric acid plays a major role in textile, food, pharmaceutical, metal and chemical industries. Melting citric acid monohydrate can give rise to itaconic anhydride. The crystals of citric acid monohydrate has the ability to preserve water up to 56 degree celsius.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Citric Acid (2014)
A switchable peroxidase mimic derived from the reversible co-assembly of cytochrome c and carbon dots
Essner JB, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 4(12), 2163-2170 (2016)
N-Glycan matrix-assisted laser desorption/ionization mass spectrometry imaging protocol for formalin-fixed paraffin-embedded tissues
Briggs MT, et al.
Rapid Communications in Mass Spectrometry, 31(10), 825-841 (2017)
An investigation into the microflora of heroin
McLauchlin J, et al.
Journal of Medical Microbiology, 51(11), 1001-1008 (2002)
Karolina Hus et al.
Frontiers in plant science, 11, 614-614 (2020-06-09)
The CRISPR/Cas9 system enables precise genome editing and is a useful tool for functional genomic studies. Here we report a detailed protocol for targeted genome editing in the model grass Brachypodium distachyon and its allotetraploid relative B. hybridum, describing gRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service