Skip to Content
Merck
All Photos(1)

Key Documents

C0682800

Cefazolin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cephazolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N8O4S3
CAS Number:
Molecular Weight:
454.51
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefazolin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(nnc1C)SCC2=C(N3[C@H](SC2)[C@@H](C3=O)NC(=O)C[n]4nnnc4)C(=O)O

InChI

1S/C14H14N8O4S3/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26)/t9-,12-/m1/s1

InChI key

MLYYVTUWGNIJIB-BXKDBHETSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefazolin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagaraju Konda et al.
Optometry and vision science : official publication of the American Academy of Optometry, 91(1), 47-53 (2013-11-12)
To investigate the utility of additional microbial analyses to detect the causative microorganism of microbial keratitis and to determine the strength of association between clinical variables. This retrospective study included 125 eyes of 123 people presenting with presumed infectious keratitis
Cheng-Kun Wu et al.
PloS one, 8(4), e61666-e61666 (2013-05-01)
Antibiotic prophylaxis with norfloxacin, intravenous ciprofloxacin, or ceftriaxone has been recommended for cirrhotic patients with gastrointestinal hemorrhage but little is known about intravenous cefazolin. This study aimed to compare the outcome of intravenous cefazolin and ceftriaxone as prophylactic antibiotics among
Alexander B A Vonk et al.
Transfusion, 54(10 Pt 2), 2608-2616 (2014-01-01)
This retrospective analysis describes blood conservation strategies and overall consumption of red blood cells (RBCs), fresh-frozen plasma (FFP), and platelet (PLT) concentrates during nonaortic cardiac surgery with cardiopulmonary bypass (CPB) in a tertiary hospital over a 10-year period. Study variables
Amira A Bhalodi et al.
Antimicrobial agents and chemotherapy, 57(11), 5679-5683 (2013-09-18)
Cefazolin, a first-generation cephalosporin with activity against methicillin-susceptible Staphylococcus aureus and streptococci, is often used to treat lower limb infections caused by these pathogens. Antimicrobial penetration is often limited in these patients due to compromised vasculature. Therefore, we sought to
Mack W Savage et al.
American journal of obstetrics and gynecology, 209(2), 108-108 (2013-05-29)
To identify risk factors for and outcomes of surgical site infections and cellulitis after abdominal hysterectomies. We used logistic regression analysis to analyze data from a case-control study of 1104 patients undergoing abdominal hysterectomies at a university hospital between Jan.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service