Skip to Content
Merck
All Photos(1)

Key Documents

C0460000

Carbidopa

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O4 · H2O
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carbidopa

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.C[C@@](Cc1ccc(O)c(O)c1)(NN)C(O)=O

InChI

1S/C10H14N2O4.H2O/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6;/h2-4,12-14H,5,11H2,1H3,(H,15,16);1H2/t10-;/m0./s1

InChI key

QTAOMKOIBXZKND-PPHPATTJSA-N

Gene Information

human ... DDC(1644)

Looking for similar products? Visit Product Comparison Guide

General description

Carbidopa is a dopaminergic drug, which is known to inhibit the conversion of dopa to dopamine. It is used in combination with levodopa for the effective treatment of restless legs syndrome and periodic leg movements in sleep.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbidopa EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The effect of carbidopa administration on urinary sodium excretion in man. Is dopamine an intrarenal natriuretic hormone?
Ball.G.S and Lee.R.M
British Journal of Clinical Pharmacology, 4(2), 115- 119 (1977)
Àlex Pericas et al.
Organic letters, 15(7), 1448-1451 (2013-03-13)
A stereoselective synthesis of L-carbidopa in seven steps and 50% overall yield from commercial compounds is described. The key step involves a highly enantioselective α-amination reaction of an acyclic β-ketoester with di-tert-butyl azodicarboxylate induced by europium and (R,R)-diphenyl-pybox.
Augmentation of the restless legs syndrome with carbidopa/levodopa
Allen.PR and Earley.JC
Sleep, 19(3), 205-213 (1996)
Robert A Hauser
Expert review of neurotherapeutics, 12(2), 133-140 (2012-02-01)
Levodopa (LD) remains the most effective medication to treat Parkinson's disease (PD). However, most patients develop motor fluctuations and dyskinesias with long-term LD use. It is thought that these response complications are due, at least in part, to LD's short
J T Hutton et al.
Neurology, 42(1 Suppl 1), 51-56 (1992-01-01)
Parkinson's disease patients treated chronically with levodopa often develop fluctuations in motor response. Motor fluctuations can be attributed in part to oscillating plasma levodopa concentrations. A controlled-release formulation containing 200 mg of levodopa and 50 mg of carbidopa provides superior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service