Skip to Content
Merck
All Photos(1)

Documents

C0166000

Calcifediol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

25-Hydroxyvitamin D3 monohydrate, 25-Hydroxycholecalciferol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2 · H2O
CAS Number:
Molecular Weight:
418.65
Beilstein:
4270041
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

sheep wool (grease)

grade

pharmaceutical primary standard

Agency

EP

API family

calcifediol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]1(CC[C@@]2([H])C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-DTOXIADCSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.Calcifediol is a biologically active metabolite of vitamin D. It is more hydrophilic and better absorbed in the intestine when compared to ergocalciferol or cholecalciferol due to the high affinity for the vitamin-D-binding protein. The drug is known to treat diseases associated with decreased intestinal absorption and obesity.

Application

Calcifediol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypovitaminosis D: is it time to consider the use of calcifediol?
Cesareo R, et al.
Nutrients, 11(5), 1016-1016 (2019)
Johanna Breuer et al.
Annals of neurology, 75(5), 739-758 (2014-04-29)
Environmental conditions (eg, latitude) play a critical role in the susceptibility and severity of many autoimmune disorders, including multiple sclerosis (MS). Here, we investigated the mechanisms underlying the beneficial effects of immune regulatory processes induced in the skin by moderate
Ting-I Lee et al.
International journal of cardiology, 176(1), 151-157 (2014-07-27)
High free fatty acid with reduced glucose utilization in diabetes mellitus (DM) impairs cardiac function. Peroxisome proliferator-activated receptors (PPARs) modulate myocardial lipid and glucose homeostasis. The active 1,25-dihydroxyvitamin D3 (1,25(OH)2D3) regulates oxidative stress and inflammation, which may play a key
Stuart M Sprague et al.
American journal of nephrology, 40(6), 535-545 (2015-01-13)
Vitamin D insufficiency drives secondary hyperparathyroidism (SHPT) and is associated with increased cardiovascular mortality in patients with chronic kidney disease (CKD). SHPT is poorly addressed by current vitamin D repletion options. The present study evaluated a novel investigational vitamin D
Kristyna Brabnikova-Maresova et al.
Arthritis research & therapy, 16(5), 460-460 (2014-10-05)
Juvenile idiopathic arthritis (JIA) is a disease associated with loss of bone mass, deterioration in bone mass quality and an increased risk of fractures. The objective of this study was to evaluate factors that predict bone mineral density (BMD) alterations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service