BP828
N-(2,6-Dichlorophenyl)-2-indolinone
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, NSC 621845
About This Item
Recommended Products
grade
pharmaceutical primary standard
description
API family: aceclofenac
API family
aceclofenac, diclofenac
form
solid
quality
British Pharmacopoeia (BP) Reference Standard
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
ClC1=C(N2C(CC3=C2C=CC=C3)=O)C(Cl)=CC=C1
InChI
1S/C14H9Cl2NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(17)18/h1-7H,8H2
InChI key
JCICIFOYVSPMHG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Also used in monographs such as:
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service