BCR402
White clover (trace elements)
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
environmental
format
matrix material
Analysis Note
For more information please see:
BCR402
BCR402
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Trace Elements: Arsenic (As), Cobalt (Co), Molybdenum (Mo), Selenium (Se)
Matrix Group: Botanicals
Trace Elements: Arsenic (As), Cobalt (Co), Molybdenum (Mo), Selenium (Se)
Matrix Group: Botanicals
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Maria Gabriela Dantas Bereta Lanza et al.
Ecotoxicology and environmental safety, 207, 111216-111216 (2020-09-12)
Low concentrations of selenium (Se) are beneficial for plant growth. Foliar Se application at high concentrations is toxic to plants due to the formation of reactive oxygen species (ROS). This study characterized Se toxicity symptoms using X-ray fluorescence (XRF) technique
André Rodrigues Dos Reis et al.
Ecotoxicology and environmental safety, 190, 110147-110147 (2020-01-10)
Selenium (Se) is an essential element for human and animal, although considered beneficial to higher plants. Selenium application at high concentration to plants can cause toxicity decreasing the physiological quality of seeds. This study aimed to characterize the Se toxicity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service