Skip to Content
Merck
All Photos(2)

Key Documents

B8675

Sigma-Aldrich

Barium sulfate

meets USP testing specifications

Synonym(s):

Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
>97.5%
form:
powder

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

>97.5%

form

powder

application(s)

pharmaceutical (small molecule)

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Light-Emitting-Diode-Induced Fluorescence from Organic Dyes for Application in Excitation-Emission Fluorescence Spectroscopy for Food System Analysis.: This study leverages the use of barium sulfate as a component in fluorescence spectroscopy for analyzing food systems. The research demonstrates how barium sulfate can enhance the fluorescence properties of organic dyes, making it a valuable tool in food safety and quality control (Vladev et al., 2024).

  • Analytical methods for quantitating sulfate in plasma and serum.: This article outlines new analytical techniques for quantifying sulfate in biological samples, with barium sulfate playing a crucial role in the precipitation and measurement processes. These methods improve the accuracy and reliability of sulfate quantitation in clinical and biochemical research (Vijayakumar and Dawson, 2024).

  • Comparison of micro-CT image enhancement after use of different vascular casting agents.: This study compares various vascular casting agents, including barium sulfate, to enhance micro-CT imaging. The findings indicate that barium sulfate significantly improves the visualization of vascular structures, making it a superior choice for detailed anatomical studies (Margolis et al., 2024).

  • Development of Lead-Free Radiation Shielding Material Utilizing Barium Sulfate and Magnesium Oxide as Fillers in Addition Cure Liquid Silicone Rubber.: This research presents a novel radiation shielding material that uses barium sulfate as a key filler. The study highlights the material′s effectiveness in providing radiation protection while being environmentally friendly and lead-free (Souza et al., 2023).


Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joon-Sung Kim et al.
American journal of physical medicine & rehabilitation, 92(10), 912-919 (2013-06-07)
The aim of this study was to report the characteristics of dysphagia in children with cerebral palsy (CP), related to gross motor function. Videofluoroscopic swallow study was performed in 29 children with CP, according to the manual of Logemann. Five
Dewey C Scheid et al.
Journal of the American Board of Family Medicine : JABFM, 26(5), 498-507 (2013-09-06)
The purpose of this study was to describe colorectal cancer screening (CRCS) practices across a variety of primary care clinics and identify the methods used by primary care physicians (PCPs) with higher rates of CRCS ("exemplars"). Physician questionnaires, structured interviews
Tafadzwa P Makarawo et al.
The American surgeon, 79(7), 728-733 (2013-07-03)
Water as an intraluminal negative contrast medium produces improved image quality with reduced artefact. However, rapid absorption of oral water in the bowel relative to speed and timing of image capturing has limited its clinical application. These findings predate advances
Zhiyue Lin et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(2), G158-G163 (2014-05-24)
This study aimed to develop and validate a method to measure bolus flow time (BFT) through the esophagogastric junction (EGJ) using a high-resolution impedance-manometry (HRIM) sleeve. Ten healthy subjects were studied with concurrent HRIM and videofluoroscopy; another 15 controls were
Christof M Sommer et al.
Investigative radiology, 48(4), 213-222 (2013-02-13)
The objective of this study was to test multimodal visibility (radiography, computed tomography [CT], and magnetic resonance imaging [MRI]) of microspheres for transarterial embolization in porcine kidneys. Currently available embolization particles (microspheres) were modified. A dense x-ray material (barium sulfate)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service