Skip to Content
Merck
All Photos(1)

Key Documents

B1158000

Bumetanide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(Aminosulfonyl)-5-(butylamino)-4-phenoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O5S
CAS Number:
Molecular Weight:
364.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bumetanide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCNc1cc(cc(c1Oc2ccccc2)S(N)(=O)=O)C(O)=O

InChI

1S/C17H20N2O5S/c1-2-3-9-19-14-10-12(17(20)21)11-15(25(18,22)23)16(14)24-13-7-5-4-6-8-13/h4-8,10-11,19H,2-3,9H2,1H3,(H,20,21)(H2,18,22,23)

InChI key

MAEIEVLCKWDQJH-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bumetanide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Flamenbaum et al.
Pharmacotherapy, 2(4), 213-222 (1982-07-01)
Bumetanide is a recently developed natriuretic and diuretic agent, belonging to the "loop" class of diuretics. Since it is rapidly and almost completely absorbed after oral administration, oral and parenteral formulations have a similar pharmacokinetic profile. Peak plasma levels are
Kristopher T Kahle et al.
Neurosurgical focus, 25(3), E22-E22 (2008-09-02)
Seizures that occur during the neonatal period do so with a greater frequency than at any other age, have profound consequences for cognitive and motor development, and are difficult to treat with the existing series of antiepileptic drugs. During development
A Ward et al.
Drugs, 28(5), 426-464 (1984-11-01)
Bumetanide is a potent 'loop' diuretic for the treatment of oedema associated with congestive heart failure, hepatic and renal diseases, acute pulmonary congestion and premenstrual syndrome and in forced diuresis during and after surgery. Bumetanide may be given orally, intravenously
C E Halstenson et al.
Drug intelligence & clinical pharmacy, 17(11), 786-797 (1983-11-01)
Bumetanide is a recently introduced diuretic that inhibits sodium transport in the thick ascending limb of the loop of Henle. It is structurally and pharmacologically similar to furosemide, but is approximately 40 times as potent on a milligram-for-milligram basis. After
D C Brater
The American journal of cardiology, 57(2), 20A-25A (1986-01-24)
Bumetanide and furosemide are potent loop diuretics; the former is 40 to 50 times more potent than the latter on a weight basis. Bumetanide is absorbed more quickly than furosemide and is twice as bioavailable. Both drugs exhibit changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service