Skip to Content
Merck
All Photos(4)

Documents

ALD00604

Sigma-Aldrich

(+)-PSI Reagent

95%

Synonym(s):

(+)-Ψ, (2R,3aR,6S,7aR)-3a-Methyl-2-((perfluorophenyl)thio)-6-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulfide, Phosphorus-Sulfur Incorporation Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16F5OPS3
CAS Number:
Molecular Weight:
446.46
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

95%

form

powder or crystals

mp

102-107 °C

Application

Air- and moisture-tolerant, this (+)-PSI Reagent was developed in the Baran lab for stereocontrolled synthesis of phosphorothioate oligonucleotides. The enantiomer, (−)-PSI Reagent, is also available (cat# ALD00602).
The PSI Reagents open access to thiophosphate-based nucleotide molecules with therapeutic promise as demonstrated by the Baran lab in the preparation of pure stereoisomers of cyclic dinucleotides (CDNs) and antisense oligonucleotides (ASOs).

Learn more about PI and PSI reagents

Legal Information

PCT/US2019/027256; U.S. Nonprovisional Patent Application #16/382,692

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle W Knouse et al.
Science (New York, N.Y.), 361(6408), 1234-1238 (2018-08-04)
Phosphorothioate nucleotides have emerged as powerful pharmacological substitutes of their native phosphodiester analogs with important translational applications in antisense oligonucleotide (ASO) therapeutics and cyclic dinucleotide (CDN) synthesis. Stereocontrolled installation of this chiral motif has long been hampered by the systemic

Articles

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service