Skip to Content
Merck
All Photos(1)

Key Documents

A9913

Supelco

Amyloglucosidase solution from Aspergillus niger

for use in Total Dietary Fiber Assay, TDF-100A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

form

liquid

Quality Level

quality

for use in Total Dietary Fiber Assay, TDF-100A

technique(s)

photometry: suitable

color

clear light yellow to amber

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application


  • Colourimetric and fluorometric substrates for measurement of pullulanase activity.: This research utilizes amyloglucosidase in developing colourimetric and fluorometric substrates for pullulanase activity measurement, offering valuable tools for analytical biochemistry applications (McCleary et al., 2014).

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Peixoto et al.
Journal of animal physiology and animal nutrition, 102(1), e111-e121 (2017-04-27)
The effects of resistant starch (RS) intake on nutrient digestibility, microbial fermentation products, faecal IgA, faecal pH, and histological features of the intestinal mucosa of old dogs were evaluated. The same formulation was extruded in two different conditions: one to
Nani Ratnaningsih et al.
International journal of biological macromolecules, 142, 191-200 (2019-09-16)
Recently, legumes starch were studied extensively due to high amylose and resistant starch contents, and low glycemic index (GI). We evaluated the impact of autoclaving-cooling cycles (single, triple, five) on the physicochemical properties, in vitro starch digestibility, and estimated glycemic
Derong Lin et al.
Journal of food science and technology, 57(11), 4247-4256 (2020-10-20)
The present study was conducted to evaluate the anti-hyperlipidemia ability of the dietary fiber extracted from okara in mice fed a high cholesterol diet. The dietary fiber was extracted from okara by combining fermentation with dynamic high-pressure microfluidization. An animal
Su-Yeon You et al.
International journal of biological macromolecules, 82, 863-870 (2015-11-20)
The in vitro digestibility as well as the molecular and crystalline structures of waxy rice starches isolated from brown rice, germinated brown rice (GBR), ultrasonicated GBR, and heat-moisture treated GBR were investigated. The germinated brown rice starch (GBRS) had a
Yong Zhuo et al.
Journal of animal science and biotechnology, 11, 47-47 (2020-05-20)
This study aimed to investigate the impacts of guar gum and cellulose as the source of dietary fiber during gestation on the reproductive performance of sows. A total of 210 sows (parities 3-6) were randomly allocated into six diets (n = 35)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service