Skip to Content
Merck
All Photos(1)

Key Documents

A7455

Sigma-Aldrich

Ammonium nitrate

BioXtra, ≥99.5%

Synonym(s):

NH4NO3 , Nitrate of ammonia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
grade:
for analytical purposes
form:
solid
solubility:
H2O: 1 M, clear, colorless

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.5%

form

solid

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

ign. residue

≤0.01%

bp

210 °C (lit.)

mp

169 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium nitrate (AN) is used as a commercial fertilizer. It is also used as a component of explosives.

Application

Ammonium nitrate is used as a component of impregnation solution in Gallyas silver stain technique for tissues.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The intercalated nuclear complex of the primate amygdala
Zikopoulos B, et al.
Neuroscience, 330, 267-290 (2016)
Opposite development of short-and long-range anterior cingulate pathways in autism
Zikopoulos B, et al.
Acta Neuropathologica, 136(5), 759-778 (2018)
Postnatal development and maturation of layer 1 in the lateral prefrontal cortex and its disruption in autism
Trutzer IM, et al.
Acta Neuropathologica Communications, 7(1), 40-40 (2019)
Coordination Compounds and Inorganics
Handbook of Thermal Analysis and Calorimetry, 5, 439-502 (2008)
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service