Skip to Content
Merck
All Photos(1)

Documents

95290

Sigma-Aldrich

Methyl Blue

for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)

Synonym(s):

Acid blue 93, Aniline blue water soluble, Cotton blue, Poirriers blue, Water blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H27N3Na2O9S3
CAS Number:
Molecular Weight:
799.80
Colour Index Number:
42780
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy (Bot., Hist.)
indicator (pH 9.4-14.0)

Quality Level

form

powder or crystals

pH

9.4-14

solubility

water: 0.01 g/10 mL, blue to very deep blue

εmax

≥200 at 310-320 nm
≥400 at 595-605 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].OS(=O)(=O)c1ccc(Nc2ccc(cc2)\C(c3ccc(Nc4ccc(cc4)S([O-])(=O)=O)cc3)=C5/C=CC(\C=C5)=N/c6ccc(cc6)S([O-])(=O)=O)cc1

InChI

1S/C37H29N3O9S3.2Na/c41-50(42,43)34-19-13-31(14-20-34)38-28-7-1-25(2-8-28)37(26-3-9-29(10-4-26)39-32-15-21-35(22-16-32)51(44,45)46)27-5-11-30(12-6-27)40-33-17-23-36(24-18-33)52(47,48)49;;/h1-24,38-39H,(H,41,42,43)(H,44,45,46)(H,47,48,49);;/q;2*+1/p-2

InChI key

MCPLVIGCWWTHFH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Blue (C.l. 42780) is a triaminotriphenylmethane dye with each amino group carrying a monosulfonated phenyl substituent. It is also known as Water blue, Acid blue 93, Cotton blue, Poirriers blue, and Aniline blue water soluble.

Application

Methyl Blue has been used:
  • to stain leaf disks for callose visualization within the cells
  • to incubate cells for 4′,6-diamidino-2-phenylindole (DAPI)/septa staining
  • as a marker for β-(1-3)-d-glucans or acidic phloroglucinol for the detection of callose and lignin
  • in Mann′s stain as it involves a methyl blue-eosin mixture

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

>572.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 300 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Guo et al.
Journal of hazardous materials, 225-226, 63-73 (2012-05-23)
Fe nanoparticles@graphene composites (FGC) are successfully synthesized by using graphene oxide (GO) as a supporting matrix. GO is first treated with Fe(3+) to form Fe(3+)@GO complexes. Then, by adding NaBH(4) solution, Fe(3+) and GO are simultaneously reduced in situ to
Ja-An Annie Ho et al.
Analytical and bioanalytical chemistry, 391(2), 479-485 (2008-02-15)
Salmonellae are ubiquitous human pathogens, which pose a danger to the elderly and children. Due to the increased number of outbreaks of human illness associated with the consumption of contaminated products in the USA and many other countries, there is
C Sahoo et al.
Journal of hazardous materials, 215-216, 302-310 (2012-03-21)
Photocatalytic degradation of methyl blue (MYB) was studied using Ag(+) doped TiO(2) under UV irradiation in a batch reactor. Catalytic dose, initial concentration of dye and pH of the reaction mixture were found to influence the degradation process most. The
Christos Haritoglou et al.
Retina (Philadelphia, Pa.), 29(9), 1266-1273 (2009-11-26)
The purpose of this study was to investigate the biocompatibility of aniline and methyl blue in a well-established cell culture model and assess the staining properties of these dyes at the level of the internal limiting membrane (ILM) in human
A Arunarani et al.
Colloids and surfaces. B, Biointerfaces, 102, 379-384 (2012-09-27)
Basic Violet 3 and Acid Blue 93 are the most important group of synthetic colourants extensively used in textile industries for dyeing cotton, wool, silk and nylon. Release of these dye pollutants in to the environment adversely affects the human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service