Skip to Content
Merck
All Photos(1)

Key Documents

94823

Supelco

Isobutyl acetate

analytical standard

Synonym(s):

Acetic acid isobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741909
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

>4 (vs air)

vapor pressure

15 mmHg ( 20 °C)

Assay

>99.8% (GC)

autoignition temp.

793 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.39 (lit.)
n20/D 1.390

bp

115-117 °C (lit.)

mp

−99 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)COC(C)=O

InChI

1S/C6H12O2/c1-5(2)4-8-6(3)7/h5H,4H2,1-3H3

InChI key

GJRQTCIYDGXPES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyl acetate is a chemical compound, commonly used as a solvent and flavoring agent in food products.[1]

Application

Isobutyl acetate may be used as a reference standard for the determination of isobutyl acetate in dry and sweet white wines using headspace solid-phase microextraction followed by high resolution gas chromatography technique (HS-SPME-HRGC).
Isobutyl acetate may be used as an analytical reference standard for the quantification of the analyte in wine samples,[2] aqueous essence and fresh fruit puree of musk melon [3] using different chromatography techniques.
Isobutyl acetate may be used as a reference standard for the determination of isobutyl acetate in dry and sweet white wines using headspace solid-phase microextraction followed by high resolution gas chromatography technique (HS-SPME-HRGC).[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - Pensky-Martens closed cup

Flash Point(C)

22 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study of sensor array and GC?MS: application to Madrid wines characterization
Santos.P.J, et al.
Sensors and Actuators B, Chemical, 102(2), 299-307 (2004)
A Comprehensive Guide to the Hazardous Properties of Chemical Substances, 49(3), 5929-5933 (2007)
Determination of esters in dry and sweet white wines by headspace solid-phase microextraction and gas chromatography
Rodriguez-Bencomo J.J, et al.
Journal of Chromatography A, 963, 213-223 (2002)
Volatile components in aqueous essence and fresh fruit of Cucumis melo cv. Athena (muskmelon) by GC-MS and GC-O.
Jordan MJ, et al.
Journal of Agricultural and Food Chemistry, 49(12), 5929-5933 (2001)
Bayram Yilmaz et al.
Reproductive toxicology (Elmsford, N.Y.), 22(4), 791-796 (2006-09-19)
Occupational exposure and sniffing of toluene-based organic solvents is an important public health problem. In this study, we have investigated the effects of paint thinner inhalation on testosterone synthesis and secretion in the male rat. A control group inhaled normal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service