Skip to Content
Merck
All Photos(1)

Key Documents

94752

Sigma-Aldrich

Vanillin

tested according to Ph. Eur.

Synonym(s):

Vanillinum, 4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
Beilstein:
472792
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

Assay

99.0-101.0% dry basis

form

powder or crystals

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

solubility

water: slightly soluble 10 g/L at 25 °C

application(s)

hematology
histology
pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Lin Lee et al.
Journal of natural products, 72(9), 1568-1572 (2009-08-21)
Two new sesquiterpene coumarins, designated 5'-acetoxy-8'-hydroxyumbelliprenin (1) and 10'R-acetoxy-11'-hydroxyumbelliprenin (2), and a new diterpene, 15-hydroxy-6-en-dehydroabietic acid (3), along with 27 known compounds, were isolated from a CHCl(3)-soluble extract of Ferula assa-foetida through bioassay-guided fractionation. The structures of the new metabolites
Igor A O Reis et al.
Food chemistry, 135(4), 2453-2461 (2012-09-18)
A single-step selective separation of two food additives was investigated using alcohol-salt aqueous two-phase systems (ATPS). The selective partitioning of two of the most used additives from a processed food waste material, vanillin and l-ascorbic acid, was successfully accomplished. The
Sartaj Tabassum et al.
European journal of medicinal chemistry, 60, 216-232 (2013-01-08)
SOD mimics with varying coligand are momentous in developing potential chemotherapeutic drugs. Cu(II) based SOD mimics 1-4 [CuLH(OAc)(H(2)O)Y)] (LH = 2-((E)-(1,3-dihydroxy-2-methylpropan-2-ylimino)methyl)-6-methoxyphenol, OAc = CH(3)COO, 1: Y = H(2)O; 2: Y = phen (1,10-phenanthroline), 3: Y = tpimH (2,4,5-triphenylimidazole); 4: Y
Yavuz Yardım et al.
Food chemistry, 141(3), 1821-1827 (2013-07-23)
A method for the determination of food additive vanillin was developed by adsorptive stripping voltammetry. Its determination was carried out at the anodically pre-treated boron-doped diamond electrode in aqueous solutions. Using square-wave stripping mode, the compound yielded a well-defined voltammetric
Ana Rita Brochado et al.
Biotechnology and bioengineering, 110(2), 656-659 (2012-09-26)
Overproduction of a desired metabolite is often achieved via manipulation of the pathway directly leading to the product or through engineering of distant nodes within the metabolic network. Empirical examples illustrating the combined effect of these local and global strategies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service