Skip to Content
Merck
All Photos(1)

Documents

92504

Supelco

Scandium Standard for ICP

TraceCERT®, 10 g/L Sc in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

10 g/L Sc in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate. Information about trace impurities are stated on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

10 g/L Sc in 5% nitric acid, prepared with high purity Sc2O3 and HNO3

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O V Anishchenko et al.
Plant physiology and biochemistry : PPB, 154, 328-340 (2020-06-28)
Accumulation of essential/beneficial and non-essential chemical elements by submerged (Stuckenia pectinata (L.) Börner) and emergent (Phragmites australis (Cav.) Trin. ex Steud.) rooted macrophytes in three water bodies was studied as dependent on water salinity (specific conductivity) and main environmental factors
Natalia Oskina et al.
Bulletin of environmental contamination and toxicology, 102(3), 335-340 (2018-10-26)
The research aimed to determine critical concentrations of heavy metals at which survival of resting eggs of the cladoceran Moina macrocopa is negatively affected. Resting eggs' viability was not affected over a 30-days exposure towards copper, cadmium, zinc or nickel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service