Skip to Content
Merck
All Photos(1)

Key Documents

91582

Supelco

Myristoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxotetradecyl)oxy]-1-propanaminium inner salt, L-Carnitine tetradecanoyl ester, C14-Carnitine, Tetradecanoyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H41NO4
CAS Number:
Molecular Weight:
371.55
Beilstein:
4149853
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -18.0±2.0°, c = 1 in methanol

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C21H41NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4/h19H,5-18H2,1-4H3/t19-/m1/s1

InChI key

PSHXNVGSVNEJBD-LJQANCHMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myristoyl-L-carnitine belongs to the acyl-L-carnitine class of compounds,[1] which are acetylated forms of L-carnitine, derived from the breakdown of amino and fatty acids.[2] The compounds are reported to be used as biomarkers in the diagnosis of carnitine deficiency.[3]

Application

Myristoyl-L-carnitine may be used as an analytical standard to determine the content of the analyte in maize plants using high-performance liquid chromatography coupled to triple quadrupole-LC MS/MS. The exogenous application of carnitine stimulates the transportation of fatty acids into mitochondria, and the possible effect of carnitine on the lipid trafficking between cytoplasm, mitochondrial matrix and the mitochondrial respiration in the leaves of maize grown under normal and cold conditions is investigated.[4]
It may be used as an analytical reference standard in the following:
  • Separation and identification of underivatized myristoyl-L-carnitine in human plasma samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).[1]
  • Determination of the analyte in dried blood samples using high-performance liquid chromatography (HPLC) coupled with electrospray ionization-tandem mass spectrometry (ESI-MS/MS).[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Acylcarnitine profiling by low-resolution LC-MS
Meierhofer D , et al.
PLoS ONE, 14(8), The-The (2019)
Exogenous carnitine application augments transport of fatty acids into mitochondria and stimulates mitochondrial respiration in maize seedlings grown under normal and cold conditions
Turk H, et al.
Cryobiology, 91, 97-103 (2019)
Ralph Fingerhut et al.
Analytical chemistry, 81(9), 3571-3575 (2009-03-28)
Electrospray ionization-tandem mass spectrometry (ESI-MS/MS) is increasingly used in newborn screening programs. Acylcarnitine profiles from dried blood spots (DBS) are used to detect fatty acid oxidation disorders, carnitine cycle disorders, and organic acidurias. Stored dried blood is also a valuable
Minzhi Peng et al.
Journal of chromatography. A, 1319, 97-106 (2013-10-31)
A simple HPLC-MS/MS method has been established to separate and identify underivatized acylcarnitine isomers. Human plasma samples were deproteinized and concentrated. Acylcarnitines were separated on a reverse phase column and detected with triple quadrupole linear ion trap mass spectrometry. Deuterium-labeled
Matrix effect?corrected liquid chromatography/tandem mass-spectrometric method for determining acylcarnitines in human urine
Abe K, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 468, 187-194 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service