Skip to Content
Merck
All Photos(1)

Key Documents

91016

Supelco

L-Methionine sulfoximine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
180.23
Beilstein:
6175446
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

>210 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

storage temp.

20-25°C

SMILES string

CS(=N)(=O)CC[C@H](N)C(O)=O

InChI

1S/C5H12N2O3S/c1-11(7,10)3-2-4(6)5(8)9/h4,7H,2-3,6H2,1H3,(H,8,9)/t4-,11?/m0/s1

InChI key

SXTAYKAGBXMACB-DPVSGNNYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Wang et al.
PloS one, 6(3), e18233-e18233 (2011-04-13)
The membrane-proximal external region (MPER) of the HIV-1 gp41 consists of epitopes for the broadly cross-neutralizing monoclonal antibodies 2F5 and 4E10. However, antigens containing the linear sequence of these epitopes are unable to elicit potent and broad neutralizing antibody responses
Hsiu-Ni Kung et al.
PLoS genetics, 7(8), e1002229-e1002229 (2011-08-20)
Although significant variations in the metabolic profiles exist among different cells, little is understood in terms of genetic regulations of such cell type-specific metabolic phenotypes and nutrient requirements. While many cancer cells depend on exogenous glutamine for survival to justify
Evan T Brower et al.
Biochemistry, 48(4), 779-785 (2009-01-28)
The initial events of HIV-1 cell infection involve the sequential binding of the viral envelope glycoprotein gp120 to the cellular CD4 receptor and the coreceptor, usually CCR5 or CXCR4. Binding to the coreceptor triggers the chain of events that culminates
Marilynn A Larson et al.
The Biochemical journal, 462(2), 329-335 (2014-06-12)
Human BChE (butyrylcholinesterase) protects against the toxicity of organophosphorus nerve agents and pesticides. BChE purified from human plasma is limited and pathogen carry-over is a concern. Unlike the native BChE tetrameric complex with a residence time of days, rBChE (recombinant
Karmela Sobczyk et al.
Neurochemical research, 40(2), 274-283 (2014-07-30)
Hepatic encephalopathy (HE) represents a neuropsychiatric syndrome, which evolves as a consequence of a low grade cerebral edema and a concomitant oxidative/nitrosative stress response. Ephrin receptors (EphR) and their ligands (ephrins) regulate astrocytic glutamate uptake and gliotransmitter release thereby governing

Protocols

Separation of L-Methionine sulfone; L-Methionine sulfoximine

Separation of L-Methionine sulfone; L-Methionine sulfoximine; L-Methionine sulfoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service