Skip to Content
Merck
All Photos(1)

Key Documents

90974

Supelco

Eleutheroside B

analytical standard

Synonym(s):

(E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-β-D-glucopyranoside, Ilexanthin A, Ligustrin, Lilacin, Magnolenin, Methoxyconiferine, Sinapyl alcohol 4-O-glucoside, Syringin, Syringoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24O9
CAS Number:
Molecular Weight:
372.37
Beilstein:
97166
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(\C=C\CO)cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C17H24O9/c1-23-10-6-9(4-3-5-18)7-11(24-2)16(10)26-17-15(22)14(21)13(20)12(8-19)25-17/h3-4,6-7,12-15,17-22H,5,8H2,1-2H3/b4-3+/t12-,13-,14+,15-,17+/m1/s1

InChI key

QJVXKWHHAMZTBY-GCPOEHJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eleutheroside B is one of the major bioactive saponins of Eleutherococcus senticosus.

Application

Eleutheroside B may be used as a reference standard for the analysis of eleutheroside B in:
  • Rat plasma by protein precipitation with methanol and high performance liquid chromatography-electrospray ionization tandem mass spectrometry (HPLC-ESI-MS/MS).
  • Rat tissue samples by solid-phase extraction (SPE) followed by HPLC combined with photodiode array detection (PDA).
  • Acanthopanax senticosus by ionic liquids-ultrasound assisted extraction (ILUAE) followed by HPLC with ultra-violet (UV) detection.
  • Eleutherococcus senticosus Maxim. by rapid resolution liquid chromatography (RRLC) equipped with multi-wavelength UV detector.
  • Acanthopanax sessiliflorus Seem. (ASS) by ultra-performance liquid chromatography-quadrupole-time-of-flight mass spectrometry (UPLC-Q-TOF-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan-yuan Song et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 45(8), 1006-1011 (2011-03-01)
This study is to investigate the therapeutic effect of syringin on adjuvant arthritis (AA) in rats and its mechanisms. Complete Freund's adjuvant (FCA) was used to induce AA in rats. Secondary paw swelling of AA rats was measured with volume
Fang Lu et al.
Biomedical chromatography : BMC, 26(10), 1269-1275 (2012-01-26)
We elucidated the structure and metabolite profile of eleutheroside B, a component derived from the extract of Acanthopanax senticosus Harms, after oral administration of the extract in rats. Samples of rat plasma were collected and analyzed by selective high-resolution liquid
Yi-Chun Lai et al.
Natural product communications, 8(3), 363-365 (2013-05-18)
Nineteen compounds and an HPLC inseparable mixture, composed of compounds 20 and 21, were isolated from the leaves and twigs of Dendropanax dentiger (Harms ex Diels) Merr. Of these, syringin (1) is the most abundant, 6'-O-apiofuranosyl dendranthemoside A (16) is
Van Kiem Phan et al.
Fitoterapia, 81(6), 485-489 (2010-01-19)
Phytochemical investigation of the methanol extract of Tinospora cordifolia aerial parts led to the isolation of four new and seven known compounds. The structures of two new aporphine alkaloids, N-formylasimilobine 2-O-beta-D-glucopyranosyl-(1-->2)-beta-D-glucopyranoside (tinoscorside A, 1) and N-acetylasimilobine 2-O-beta-D-glucopyranosyl-(1-->2)-beta-D-glucopyranoside (tinoscorside B, 2)
Jianguo Xing et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 34(13), 1661-1664 (2009-10-31)
To study the effects of different penetration enhancers on the transcutaneous permeability of syringin, chologenic acid and rutin in Xuelian cataplasm in vitro and to determine the effective enhancer. Using improved Franz-type diffusion cell and excised big mouse skin in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service