Skip to Content
Merck
All Photos(1)

Key Documents

88561

Sigma-Aldrich

2,2′-Thiodiethanol

purum, ≥95.0% (GC)

Synonym(s):

2,2′-Thiobis(ethanol), Bis(2-hydroxyethyl) sulfide, Thiodiethylene glycol, Thiodiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(CH2CH2OH)2
CAS Number:
Molecular Weight:
122.19
Beilstein:
1236325
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.521
n20/D 1.5215 (lit.)

bp

164-166 °C/20 mmHg (lit.)

mp

−16 °C (lit.)

density

1.221 g/mL at 25 °C (lit.)

functional group

hydroxyl
thioether

SMILES string

OCCSCCO

InChI

1S/C4H10O2S/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

YODZTKMDCQEPHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Thiodiethanol is used in the preparation of a wide range of gold complexes, which include Au(I) based catalysts such as chloro(triphenylphosphine)gold(I) [Au(PPh3)Cl], chloro(dimethyl sulfide)gold(I) [Au(SMe2)Cl] and chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) [IPrAuCl]. It can also be used to build calixarene frameworks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold (I)?Catalyzed Intermolecular Cyclopropanation of Enynes with Alkenes: Trapping of Two Different Gold Carbenes.
Lopez S, et al.
Angewandte Chemie (International Edition in English), 118(36), 6175-6178 (2006)
Binding and extraction of alkali and alkaline earth metals by nano-baskets of calix [4] arene-1, 2-crown-3.
Mokhtari B and Pourabdollah K
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 73(1-4), 269-277 (2012)
Stereo-and regioselective gold-catalyzed hydroamination of internal alkynes with dialkylamines.
Hesp K D, et al.
Journal of the American Chemical Society, 132(51), 18026-18029 (2010)
A general synthesis for gold (I) complexes.
Al?Sa'Ady A K, et al.
Inorg. Synth., 23, 191-194 (1985)
Networking of calixcrowns: From heteronuclear endo/exocyclic coordination polymers to a photoluminescence switch.
Lee J Y, et al.
Journal of the American Chemical Society, 130(42), 13838-13839 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service